Skip to Content
Merck
All Photos(1)

Documents

17933

Supelco

Hydrogen chloride solution

~1.25 M in 2-propanol, for GC derivatization, LiChropur

Synonym(s):

Hydrochloric acid in 2-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCl · CH3CH(OH)CH3
Molecular Weight:
36.46
Beilstein:
1098214
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

product name

Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur

grade

for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Alkylations

concentration

~1.25 M HCl (T)

technique(s)

gas chromatography (GC): suitable

density

0.821 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diluted solutions of Hydrogen chloride in 2-propanol is used as a derivatizing agent for GC.
It may be used in measuring their conductance at 25°C.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Effect of water on proton migration in alcoholic solvents-II. conductance of hydrogen chloride in 2-propanol at 25? C.
De Lisi, R., and M. Goffredi.
Electrochimica Acta, 17, 2001-2008 (1972)
Thomas Krohn et al.
European journal of nuclear medicine and molecular imaging, 42(2), 210-214 (2014-09-25)
To determine the frequency of seemingly pathological retroperitoneal uptake in the location of the coeliac ganglia in patients undergoing [(68)Ga]PSMA-HBED PET/CT. The study included 85 men with prostate cancer referred for [(68)Ga]PSMA-HBED PET/CT. The PET/CT scans were evaluated for the
Jay G Hosking et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(4), 1005-1015 (2014-10-21)
Successful decision making often requires weighing a given option's costs against its associated benefits, an ability that appears perturbed in virtually every severe mental illness. Animal models of such cost/benefit decision making overwhelmingly implicate mesolimbic dopamine in our willingness to
Mahsa Sadeghi et al.
British journal of pharmacology, 172(2), 460-468 (2014-01-01)
Tapentadol is a novel analgesic that combines moderate μ-opioid receptor agonism and noradrenaline reuptake inhibition in a single molecule. Both mechanisms of action are involved in producing analgesia; however, the potency and efficacy of tapentadol in individual neurons has not
Ann Brinkmalm et al.
Molecular neurodegeneration, 9, 53-53 (2014-11-25)
Synaptic degeneration is an early pathogenic event in Alzheimer's disease, associated with cognitive impairment and disease progression. Cerebrospinal fluid biomarkers reflecting synaptic integrity would be highly valuable tools to monitor synaptic degeneration directly in patients. We previously showed that synaptic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service