Merck
  • Toxicokinetic and mechanistic basis for the safety and tolerability of liposomal amphotericin B.
Toxicokinetic and mechanistic basis for the safety and tolerability of liposomal amphotericin B.
Expert opinion on drug safety (2013-08-13)
Angela S Loo, Saif A Muhsin, Thomas J Walsh
ABSTRACT
Amphotericin B (AmB) was first approved by the US Food and Drug Administration in 1959 with sodium deoxycholate (DAmB, Fungizone®). Extensive toxicities associated with the drug led to the development of lipid formulations of AmB, including liposomal amphotericin B (L-AmB, AmBisome®). Phase I studies as well as comparative Phase III clinical trials indicate that L-AmB is associated with less nephrotoxicity and reduced infusion-related toxicity. There is, however, no recent comprehensive review of the safety and tolerability of L-AmB. This article reviews the safety, tolerability and the mechanisms of the major toxicities associated with L-AmB, including nephrotoxicity, infusion-related reactions (IRRs), anemia and thrombocytopenia, and hepatic abnormalities. The article further discusses the mechanism of action and pharmacokinetics of L-AmB. L-AmB is a broad-spectrum antifungal agent that has significantly reduced toxicities compared to its predecessor, DAmB.
Product Description
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Amphotericin B for microbiological assay, European Pharmacopoeia (EP) Reference Standard