Merck
  • Cadmium accumulation in gill, liver, kidney and muscle tissues of common carp, Cyprinus carpio, and Nile tilapia, Oreochromis niloticus.
Cadmium accumulation in gill, liver, kidney and muscle tissues of common carp, Cyprinus carpio, and Nile tilapia, Oreochromis niloticus.
Bulletin of environmental contamination and toxicology (2014-02-15)
Burcu Yeşilbudak, Cahit Erdem
ABSTRACT
Accumulation of cadmium in gill, liver, muscle and kidney tissues of Cyprinus carpio and Oreochromis niloticus were investigated in fish exposed to 0.5 ppm cadmium over 1, 15 and 30 days under controlled laboratory conditions. Tissue accumulation of the metal was measured using Atomic Absorption Spectrophotometric techniques. Cadmium accumulation in gill, liver, kidney and muscle, tissues of C. carpio and O. niloticus exposed to metal for 1, 15 and 30 days increased significantly compared with the control group (p < 0.05), except muscle tissue of O. niloticus. A general increase was observed in Cd accumulation with increasing exposure periods. Highest metal accumulation was observed in kidney followed by liver, gill and muscle tissues in both species. Liver accumulation of Cd was higher in C. carpio than O. niloticus, whereas kidney accumulation of the metal was higher in O. niloticus than C. carpio.
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cadmium, foil, not light tested, 25x25mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.002mm, 99.7+%
Cadmium, foil, 50mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, rod, 100mm, diameter 5.0mm, as drawn, 99.99+%
Cadmium, foil, light tested, 50x50mm, thickness 0.025mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.025mm, 99.7+%
Cadmium, foil, 50mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.025mm, 99.7+%
Cadmium, foil, light tested, 100x100mm, thickness 0.025mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.005mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 50mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 8mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, tube, 100mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 8mm disks, thickness 0.01mm, 99.7+%
Cadmium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, wire reel, 0.1m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.015mm, 99.7+%
Cadmium, tube, 200mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, light tested, 50x50mm, thickness 0.01mm, 99.7+%