Merck
  • Haemocyte apoptosis of the tiger shrimp Penaeus monodon exposed to cadmium.
Haemocyte apoptosis of the tiger shrimp Penaeus monodon exposed to cadmium.
Bulletin of environmental contamination and toxicology (2014-01-01)
Jian-An Xian, Bin Li, Hui Guo, Yu-Tao Miao, Jian-Min Ye, Li-Na Feng, Chao-Xia Ye, Xun-Bin Pan, An-Li Wang, Xuan-Ming Hao
ABSTRACT
This study investigated the effect of ambient Cadmium (Cd) on haemocyte apoptosis of the shrimp, Penaeus monodon. Cellular response was determined in Cd-exposed (0, 0.05, 0.5 and 5 mg L(-1)) shrimp. Results showed that 0.05 mg L(-1) Cd(2+) had no significant effect on the haemocyte parameters during the 48 h exposure. Cadmium at doses of 0.5 and 5 mg L(-1) depressed the total haemocyte count (THC), and increased reactive oxygen species (ROS) production and apoptosis ratio in haemocytes. Esterase activity increased in shrimp exposed to 0.5 mg L(-1) Cd(2+) for 6 h, and decreased to the initial level later. Depressed esterase activity could be observed in shrimp after 24 and 48 h exposure to 5 mg L(-1) Cd(2+). These results demonstrated that Cd(2+) modified esterase activity and induced ROS generation, which led to haemocyte apoptosis and THC reduction. Oxidative stress is one of the induction mechanisms for Cd-caused apoptosis of shrimp haemocytes.
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cadmium, foil, not light tested, 25x25mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.002mm, 99.7+%
Cadmium, foil, 50mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, rod, 100mm, diameter 5.0mm, as drawn, 99.99+%
Cadmium, foil, light tested, 50x50mm, thickness 0.025mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.025mm, 99.7+%
Cadmium, foil, 50mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.025mm, 99.7+%
Cadmium, foil, light tested, 100x100mm, thickness 0.025mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.005mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 50mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 8mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, tube, 100mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 8mm disks, thickness 0.01mm, 99.7+%
Cadmium, foil, 4mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, wire reel, 0.1m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.015mm, 99.7+%
Cadmium, tube, 200mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, light tested, 50x50mm, thickness 0.01mm, 99.7+%