Merck
  • 1,5-anhydroglucitol and postprandial hyperglycemia as measured by continuous glucose monitoring system in moderately controlled patients with diabetes.
1,5-anhydroglucitol and postprandial hyperglycemia as measured by continuous glucose monitoring system in moderately controlled patients with diabetes.
Diabetes care (2006-05-30)
Kathleen M Dungan, John B Buse, Joseph Largay, Mary M Kelly, Eric A Button, Shuhei Kato, Steven Wittlin
ABSTRACT
Postprandial hyperglycemia is often inadequately assessed in diabetes management. Serum 1,5-anhydroglucitol (1,5-AG) drops as serum glucose rises above the renal threshold for glucose and has been proposed as a marker for postprandial hyperglycemia. The objective of this study is to demonstrate the relationship between 1,5-AG and postprandial hyperglycemia, as assessed by the continuous glucose monitoring system (CGMS) in suboptimally controlled patients with diabetes. Patients with type 1 or type 2 diabetes and an HbA(1c) (A1C) between 6.5 and 8% with stable glycemic control were recruited from two sites. A CGMS monitor was worn for two consecutive 72-h periods. Mean glucose, mean postmeal maximum glucose (MPMG), and area under the curve for glucose above 180 mg/dl (AUC-180), were compared with 1,5-AG, fructosamine (FA), and A1C at baseline, day 4, and day 7. 1,5-AG varied considerably between patients (6.5 +/- 3.2 mug/ml [means +/- SD]) despite similar A1C (7.3 +/- 0.5%). Mean 1,5-AG (r = -0.45, P = 0.006) correlated with AUC-180 more robustly than A1C (r = 0.33, P = 0.057) or FA (r = 0.38, P = 0.88). MPMG correlated more strongly with 1,5-AG (r = -0.54, P = 0.004) than with A1C (r = 0.40, P = 0.03) or FA (r = 0.32, P = 0.07). 1,5-AG reflects glycemic excursions, often in the postprandial state, more robustly than A1C or FA. 1,5-AG may be useful as a complementary marker to A1C to assess glycemic control in moderately controlled patients with diabetes.
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Supelco
1-Butanol, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Levocarnitine, European Pharmacopoeia (EP) Reference Standard