Skip to Content
Merck
All Photos(3)

Key Documents

266264

Sigma-Aldrich

Chromium

powder, 99.5%, −100 mesh

Synonym(s):

Cd

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr
CAS Number:
Molecular Weight:
52.00
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.5%

form

powder

reaction suitability

core: chromium
reagent type: catalyst

resistivity

12.7 μΩ-cm, 20°C

particle size

−100 mesh

bp

2672 °C (lit.)

mp

1857 °C (lit.)

density

7.14 g/mL at 25 °C (lit.)

SMILES string

[Cr]

InChI

1S/Cr

InChI key

VYZAMTAEIAYCRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chromium powder can be used in the microwave assisted synthesis of a bis(toluene)chromium boron complex, [Cr(η-C6H5CH3)2]+ (BPh4)-. It can also be used in the solid-state synthesis of chromium sulfide.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic reactions using metal powders under microwave irradiation.
Whittaker A G and Mingos D M P
J. Chem. Soc., Dalton Trans., 21, 3967-3970 (2002)
Microwave-assisted solid-state reactions involving metal powders.
GavinaWhittaker A and Michael P aMingos D
J. Chem. Soc., Dalton Trans., 12, 2073-2079 (1995)
B Seifert et al.
Journal of exposure analysis and environmental epidemiology, 10(2), 103-114 (2000-05-03)
The German Environmental Survey (GerES) is a large-scale population study which has repeatedly been carried out in Germany. GerES I was conducted in 1985/1986 followed by GerES IIa in 1990/1991 (West Germany) and GerES IIb in 1991/1992 (East Germany). GerES
J A J Vanoirbeek et al.
Journal of toxicology and environmental health. Part A, 66(4), 393-409 (2003-01-30)
Chromium-based catalysts are used for the synthesis of polyethylene, but little is known about the hazard and biomonitoring possibilities of this type of chromium for workers who may be occupationally exposed to such compounds. Therefore, the bioavailability and toxicokinetics of
Jacob P Thyssen et al.
Contact dermatitis, 69(5), 276-279 (2013-10-15)
A patient who had suffered from persistent generalized dermatitis for 7 years was diagnosed with cobalt sensitization, and his leather couch was suspected as the culprit, owing to the clinical presentation mimicking allergic chromium dermatitis resulting from leather furniture exposure. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service