Merck
Serum 8-hydroxy-guanine levels are increased in diabetic patients.
Diabetes care (2001-04-24)
C S Shin, B S Moon, K S Park, S Y Kim, S J Park, M H Chung, H K Lee
ABSTRACT
The production of reactive oxygen species is increased in diabetic patients, especially in those will poor glycemic control. We have investigated oxidative damage in type 2 diabetic patients using serum 8-hydroxyguanine (8-OHG) as a biomarker. We studied 41 type 2 diabetic patients and compared them with 3 nondiabetic control subjects. Serum 8-OHG concentration was assayed using high-pressure liquid chromatography. The type 2 diabetic patients had significantly higher concentrations of 8-OHG in their serum than the control subjects (5.03 +/- 0.69 vs. 0.96 +/- 0.15 pmol/ml P < 0.01). There was no association between the levels of 8-OHG and HbA1c. We also could not and any correlation between serum 8-OHG levels and age, duration of diabetes, serum lipids, or creatinine or albumin exeretion rate. Creatinine clearance showed marginal correlation with serum 8-OHG levels (P = 0.06). Among the diabetic patients, those with proliferative retinopathy had significantly higher 8-OHG levels than those with nonproliferative retinopathy or without retinopathy. Likewise, the serum 8-OHG levels in patients who had advanced nephropathy (azotemia) were higher than in patients with normoalbuminuria, microalbuminuria, or overt proteinuria. Our findings show that measuring serum 8-OHG is a novel convenient method for evaluating oxidative DNA damage. Diabetic patients, especially those with advanced microvascular complications, had significantly higher serum 8-OHG levels; this suggests that such changes may contribute to the development of microvascular complications of diabetes.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Supelco
Acetone, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF