Merck
  • Evaluation of chromium, cobalt and manganese in biological samples (scalp hair, blood, and urine) of Pakistani viral hepatitis (A-E) patients and controls.
Evaluation of chromium, cobalt and manganese in biological samples (scalp hair, blood, and urine) of Pakistani viral hepatitis (A-E) patients and controls.
Clinical laboratory (2013-06-04)
Hassan Imran Afridi, Tasneem Gul Kazi, Naveed Kazi, Naeemullah, Sadaf Sadia Arain, Kapil Dev Brahman, Sham Kumar Wadhwa
ABSTRACT
The aim of the present study was to compare the level of chromium (Cr), cobalt (Co), and manganese (Mn) in biological samples (blood, urine, and scalp hair) of patients suffering from different types of viral hepatitis (A, B, C, D, and E; n = 521) of both genders, ages ranging from 31 - 45 years. For comparative study, 255 age-matched control subjects of both genders residing in the same city were selected as referents. The digests of all biological samples were analysed for Cr, Co, and Mn by electrothermal atomic absorption spectrometry (ETAAS). The validity and accuracy of the methodology was checked by using certified reference materials (CRMs) and compared with those values obtained by conventional wet acid digestion method on same CRMs. The results of this study showed that the mean values of Cr, Co, and Mn were higher in blood and scalp hair samples of hepatitis patients than in age-matched control subjects. The urinary levels of these elements were found to be higher in the hepatitis patients than in the age-matched healthy controls (p <0.001). These results are consistent with literature-reported data, confirming that the overload of these trace elements can directly cause lipid peroxidation and eventually hepatic damage.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis