Merck
Common Markers and Small Molecule Inhibitors in Golgi Studies.
Methods in molecular biology (Clifton, N.J.) (2022-12-14)
Sarah Bui, Drew Stark, Jie Li, Jianchao Zhang, Yanzhuang Wang
ABSTRACT
In this chapter, we provide a detailed guide for the application of commonly used small molecules to study Golgi structure and function in vitro. Furthermore, we have curated a concise, validated list of endomembrane markers typically used in downstream assays to examine the consequent effect on the Golgi via microscopy and western blot after drug treatment. This chapter will be useful for researchers beginning their foray into the field of intracellular trafficking and Golgi biology.
Sigma-Aldrich
Anti-α1,2-Mannosidase IA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Tunicamycin, Streptomyces lysosuperficus
Sigma-Aldrich
Bisindolylmaleimide I, A highly selective, cell-permeable, and reversible protein kinase C (PKC) inhibitor (IC₅₀ = 10 nM) that is structurally similar to staurosporine.
Sigma-Aldrich
Staurosporine, Streptomyces sp., Staurosporine, CAS 62996-74-, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of protein kinases (IC₅₀ = 7, 20, 1.3, 0.7, & 8.5 nM for PKA, CAMK, MLCK, PKC, & PKG, respectively).
Sigma-Aldrich
KN-93, ≥98% (HPLC)
Sigma-Aldrich
LY-411575, ≥97% (HPLC)
Sigma-Aldrich
γ-Secretase Inhibitor X, InSolution, ≥90%, 1 mM
Sigma-Aldrich
Latrunculin B from Latruncula magnifica, ≥80% (HPLC), solid
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Roche
Antipain dihydrochloride, crystalline, suitable for cell culture, pkg of 10 mg
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
FIBRONECTIN FROM HUMAN PLASMA, liquid, 0.1% (Solution), BioReagent, suitable for cell culture
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Epoxomicin, ≥95% (HPLC), solid
Sigma-Aldrich
MG-132, ≥95% by HPLC, Potent, reversible, and cell-permeable proteasome inhibitor (Ki = 4 nM).
Sigma-Aldrich
ML-7, powder
Sigma-Aldrich
Pyocyanin, from Pseudomonas aeruginosa, ≥98% (HPLC)
Sigma-Aldrich
TAPI-1, InSolution, ≥97%, inhibits MMPs and TACE
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
β-Secretase Inhibitor II, The β-Secretase Inhibitor II, also referenced under CAS 263563-09-3, controls the biological activity of β-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.
Sigma-Aldrich
Fucosyltransferase Inhibitor, 2F-Peracetyl-Fucose, 2F-Peracetyl-Fucose is a cell-permeable fluorinated fucose derivative that acts as an inhibitor of fucosyltransferases following its uptake and metabolic transformation into a GDP-fucose mimetic.
Millipore
BAPTA/AM, Membrane-permeable form of BAPTA.
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Roche
Chymostatin