Skip to Content
Merck
All Photos(2)

Key Documents

474787

Sigma-Aldrich

MG-132

≥95% by HPLC, Potent, reversible, and cell-permeable proteasome inhibitor (Kᵢ = 4 nM).

Synonym(s):

Z-Leu-Leu-Leu-al, Proteasome Inhibitor XI, Z-Leu-Leu-Leu-CHO, Carbobenzoxy-L-leucyl-L-leucyl-L-leucinal, MG-132

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H41N3O5
CAS Number:
Molecular Weight:
475.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

MG-132, ≥95% by HPLC, Potent, reversible, and cell-permeable proteasome inhibitor (Ki = 4 nM).

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL
ethanol: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[H]C(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)OCc1ccccc1

InChI

1S/C26H41N3O5/c1-17(2)12-21(15-30)27-24(31)22(13-18(3)4)28-25(32)23(14-19(5)6)29-26(33)34-16-20-10-8-7-9-11-20/h7-11,15,17-19,21-23H,12-14,16H2,1-6H3,(H,27,31)(H,28,32)(H,29,33)/t21-,22-,23-/m0/s1

InChI key

TZYWCYJVHRLUCT-VABKMULXSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Z-Leu-Leu-Leu-al

General description

Potent, reversible, and cell-permeable proteasome inhibitor (Ki = 4 nM). Reduces the degradation of ubiquitin-conjugated proteins in mammalian cells and permeable stains of yeast by the 26S complex without affecting its ATPase or isopeptidase activities. Activates c-Jun N-terminal kinase (JNK1) which initiates apoptosis. Inhibits NF-κB activation (IC50 = 3 µM). This material is ≥95% pure by HPLC. A material with ≥98% purity by HPLC (Cat. No. 474790) is also available. Also available as a 20 mM solution in DMSO (Cat. No. 474788).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Steinhilb, M.L., et al. 2001. J. Biol. Chem.276, 4476.
Meriin, A.B., et al. 1998. J. Biol. Chem. 273, 6373.
Adams, J., and Stein, R. 1996. Ann. Rep. Med. Chem.31, 279.
Klafki, H.W., et al. 1996. J. Biol. Chem.271, 2865.
Lee, D.H., and Goldberg, A.L., 1996. J. Biol. Chem.271, 27280.
Wiertz, E.J., et al. 1996. Cell84, 769.
Jensen, T.J., et al. 1995. Cell 83, 129.
Read, M.A., et al. 1995. Immunity2, 493.
Rock, K.L., et al. 1994. Cell 78, 761.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harihar Milaganur Mohan et al.
The Journal of biological chemistry, 295(49), 16754-16772 (2020-09-27)
α1-antitrypsin (AAT) regulates the activity of multiple proteases in the lungs and liver. A mutant of AAT (E342K) called ATZ forms polymers that are present at only low levels in the serum and induce intracellular protein inclusions, causing lung emphysema
Haiyun Chen et al.
Clinical and translational medicine, 12(4), e574-e574 (2022-04-08)
Senescence-associated pathological cardiac hypertrophy (SA-PCH) is associated with upregulation of foetal genes, fibrosis, senescence-associated secretory phenotype (SASP), cardiac dysfunction and increased morbidity and mortality. Therefore, we conducted experiments to investigate whether GATA4 accumulation induces SA-PCH, and whether Bmi-1-RING1B promotes GATA4
Tanmoy Mondal et al.
EMBO molecular medicine, 13(3), e12716-e12716 (2021-02-16)
Lack of effective immune infiltration represents a significant barrier to immunotherapy in solid tumors. Thus, solid tumor-enriched death receptor-5 (DR5) activating antibodies, which generates tumor debulking by extrinsic apoptotic cytotoxicity, remains a crucial alternate therapeutic strategy. Over past few decades
Shuiyan Wu et al.
Cancer cell international, 21(1), 230-230 (2021-04-24)
T-cell acute lymphoblastic leukemia (T-ALL) is an aggressive disease with a high risk of induction failure and poor outcomes, with relapse due to drug resistance. Recent studies show that bromodomains and extra-terminal (BET) protein inhibitors are promising anti-cancer agents. ARV-825
Ping Liu et al.
Current genetics, 66(4), 813-822 (2020-04-02)
Slowing down aging-associated accumulation of molecular damage or its prevention represents a promising therapeutic paradigm to combat aging-related disease and death. While several chemical compounds extend lifespan in model organisms, their mechanism of action is often unknown, reducing their therapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service