Merck
  • Characterization and anions inhibition studies of an α-carbonic anhydrase from the teleost fish Dicentrarchus labrax.
Characterization and anions inhibition studies of an α-carbonic anhydrase from the teleost fish Dicentrarchus labrax.
Bioorganic & medicinal chemistry (2011-01-08)
Deniz Ekinci, Saltuk Buğrahan Ceyhun, Murat Sentürk, Deryanur Erdem, Omer İrfan Küfrevioğlu, Claudiu T Supuran
ABSTRACT
Carbonic anhydrase (CA; EC 4.2.1.1) was purified from the gill of the teleost fish Dicentrarchus labrax (European seabass). The purification procedure consisted of a single step affinity chromatography on Sepharose 4B-tyrosine-sulfanilamide. The enzyme was purified 84.9-fold with a yield of 58%, and a specific activity of 838.9 U/mg proteins. It has an optimum pH at 8.0; an optimum temperature at 10°C. The kinetic parameters of this enzyme were determined for its esterase activity, with 4-nitrophenyl acetate (NPA) as substrate. The following anions, H₂NSO₃⁻, I⁻, SCN⁻, NO₃⁻, NO₂⁻, N₃⁻, Br⁻, Cl⁻, SO₄²⁻, and F⁻ showed inhibitory effects on the enzyme. Sulfamic acid, iodide, and thiocyanate exhibited the strongest inhibitory action, in the micromolar range (K(i)s of 87-187 μM). NO₃⁻, NO₂⁻ and N₃⁻ were moderate inhibitors, whereas other anions showed only weak actions. All tested anions inhibited the enzyme in a competitive manner. Our findings indicate that these anions inhibit the fish enzyme in a similar manner to other α-CAs from mammals investigated earlier, but the susceptibility to various anions differs significantly between the fish and mammalian CAs.
Sigma-Aldrich
Potassium fluoride on alumina, extent of labeling: 40 wt. % loading
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrite solution
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl