Merck
  • Inhibition of the β-carbonic anhydrase from Streptococcus pneumoniae by inorganic anions and small molecules: Toward innovative drug design of antiinfectives?
Inhibition of the β-carbonic anhydrase from Streptococcus pneumoniae by inorganic anions and small molecules: Toward innovative drug design of antiinfectives?
Bioorganic & medicinal chemistry (2010-12-18)
Peter Burghout, Daniela Vullo, Andrea Scozzafava, Peter W M Hermans, Claudiu T Supuran
ABSTRACT
The Gram-positive bacterium Streptococcus pneumoniae is a human respiratory tract pathogen that contributes significantly to global mortality and morbidity. It was recently shown that this bacterial pathogen depends on a conserved β-carbonic anhydrase (CA, EC 4.2.1.1) for in vitro growth in environmental ambient air and during intracellular survival in host cells. Hence, it is to be expected that this pneumococcal carbonic anhydrase (PCA) contributes to transmission and pathogenesis of the bacterium, making it a potential therapeutic target. In this study, purified recombinant PCA has been further characterized kinetically and for inhibition with a series of inorganic anions and small molecules useful as leads. PCA has appreciable activity as catalyst for the hydration of CO(2) to bicarbonate, with a k(cat) of 7.4×10(5)s(-1) and k(cat)/K(m) of 6.5×10(7) M(-1)s(-1) at an optimum pH of 8.4. Inorganic anions such as chloride, bromide, iodide, cyanate, selenocyanate, trithiocarbonate, and cyanide were effective inhibitors of PCA (K(I)s of 21-98μM). Sulfamide, sulfamic acid, phenylboronic, phenylarsonic acid, and diethyldithiocarbamate showed inhibition constants in the low micromolar/submicromolar range (K(I)s of 0.61-6.68μM), whereas that of the sulfonamide acetazolamide was in the nanomolar range (K(I)s 89nM). In conclusion, our results show that PCA can effectively be inhibited by a range of molecules that could be interesting leads for obtaining more potent PCA inhibitors. PCA might be a novel target for designing antimicrobial drugs with a new mechanism of action.
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Supelco
Density Standard 1251 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Sodium chloride physiological solution, BioUltra, tablet
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride physiological solution, BioUltra, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sulfamide, 99%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%