Skip to Content
Merck
All Photos(1)

Key Documents

211370

Sigma-Aldrich

Sulfamide

99%

Synonym(s):

Imidosulfamic acid, Sulfamoylamine, Sulfonyl diamide, Sulfuryl amide, Sulfuryl diamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2)2SO2
CAS Number:
Molecular Weight:
96.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

90-92 °C (lit.)

solubility

water: soluble 50 mg/mL, clear, colorless to faintly yellow

density

1.611 g/mL at 25 °C (lit.)

SMILES string

NS(N)(=O)=O

InChI

1S/H4N2O2S/c1-5(2,3)4/h(H4,1,2,3,4)

InChI key

NVBFHJWHLNUMCV-UHFFFAOYSA-N

Gene Information

human ... CA1(759) , CA2(760)

Looking for similar products? Visit Product Comparison Guide

General description

Sulfamide, a polar aprotic solvent compatible with Grignard reagents, is used as a functional group in medicinal chemistry.

Application

Sulfamide was used in the synthesis of:
  • Schiff bases of the type ArCH=NSO2NH2
  • 1H,3H-2,1,3-benzothiadiazin-4-one-2,2-dioxide (BTDD)
  • sulfamide analogs of oleoylethanolamide analogs in a study of PPARα activation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfamides and sulfonamides as polar aprotic solvents
Herman R G et al.
The Journal of Organic Chemistry, 52, 479-483 (1987)
Bacterial cleavage of nitrogen to sulfone bonds in sulfamide and 1H-2, 1, 3-benzothiadiazin-4 (3H)-one 2, 2-dioxide: formation of 2-nitrobenzamide by Gordonia sp.
Rein U and Cook AM.
FEMS Microbiology Letters, 172(2), 107-113 (1999)
Anomeric effects in sulfamides
Eric H et al.
The Journal of Physical Chemistry A, 120, 3677-3682 (2016)
Carolina Cano et al.
Journal of medicinal chemistry, 50(2), 389-393 (2007-01-19)
Long chain saturated and unsaturated alkyl sulfamide and propyl sulfamide derivatives, analogs of oleoylethanolamide, have been synthesized and evaluated in vivo and in vitro as peroxisome proliferator activated receptor alpha (PPARalpha) activators. Additionally, the anorexic effects of the new compounds
Jean-Yves Winum et al.
Bioorganic & medicinal chemistry letters, 15(9), 2353-2358 (2005-04-20)
A small library of N-hydroxysulfamides was synthesized by an original approach in order to investigate whether this zinc-binding function is efficient for the design of inhibitors targeting the cytosolic (hCA I and II) and transmembrane, tumor-associated (hCA IX and XII)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service