Skip to Content
Merck
All Photos(4)

Key Documents

900506

Sigma-Aldrich

Sodium bicarbonate

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%

Synonym(s):

Sodium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein:
4153970
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5%
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Agency

suitable for EPA 300

product line

ReagentPlus®
Redi-Dri

Assay

≥99.5%

form

powder

quality

free-flowing

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

density

2.16 g/mL at 25 °C (lit.)

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophia Zoungas et al.
Annals of internal medicine, 151(9), 631-638 (2009-11-04)
Intravenous sodium bicarbonate has been proposed to reduce the risk for contrast-induced nephropathy (CIN). To determine the effect of sodium bicarbonate on the risk for CIN. MEDLINE, PubMed, EMBASE, and the Cochrane Central Register of Controlled Trials from 1950 to
Eric A J Hoste et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 25(3), 747-758 (2009-08-26)
There have been conflicting reports on the use of intravenous administration of sodium bicarbonate for prevention of contrast-induced acute kidney injury (CI-AKI). The aim of this study was to evaluate the use of sodium bicarbonate for prevention of CI-AKI. This
Shay P McGuinness et al.
Critical care medicine, 41(7), 1599-1607 (2013-05-21)
Cardiac surgery-associated acute kidney injury occurs in up to 50% of patients and is associated with increased mortality and morbidity. This study aimed to discover if perioperative urinary alkalinization with sodium bicarbonate infusion reduces the prevalence of cardiac surgery-associated acute
Michael Haase et al.
PLoS medicine, 10(4), e1001426-e1001426 (2013-04-24)
Preliminary evidence suggests a nephroprotective effect of urinary alkalinization in patients at risk of acute kidney injury. In this study, we tested whether prophylactic bicarbonate-based infusion reduces the incidence of acute kidney injury and tubular damage in patients undergoing open
Kate M Hatzopoulos et al.
Investigative ophthalmology & visual science, 55(6), 3957-3970 (2014-06-05)
Retinal angiogenesis, neural dysfunction, and microglial activation occur in both humans with retinopathy of prematurity and in animal models of the disease. The aim of this study was to assess whether blockade of the renin-angiotensin system ameliorated these effects in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service