Merck
  • FAMIN Is a Multifunctional Purine Enzyme Enabling the Purine Nucleotide Cycle.
FAMIN Is a Multifunctional Purine Enzyme Enabling the Purine Nucleotide Cycle.
Cell (2020-01-25)
M Zaeem Cader, Rodrigo Pereira de Almeida Rodrigues, James A West, Gavin W Sewell, Muhammad N Md-Ibrahim, Stephanie Reikine, Giuseppe Sirago, Lukas W Unger, Ana Belén Iglesias-Romero, Katharina Ramshorn, Lea-Maxie Haag, Svetlana Saveljeva, Jana-Fabienne Ebel, Philip Rosenstiel, Nicole C Kaneider, James C Lee, Trevor D Lawley, Allan Bradley, Gordon Dougan, Yorgo Modis, Julian L Griffin, Arthur Kaser
ABSTRACT
Mutations in FAMIN cause arthritis and inflammatory bowel disease in early childhood, and a common genetic variant increases the risk for Crohn's disease and leprosy. We developed an unbiased liquid chromatography-mass spectrometry screen for enzymatic activity of this orphan protein. We report that FAMIN phosphorolytically cleaves adenosine into adenine and ribose-1-phosphate. Such activity was considered absent from eukaryotic metabolism. FAMIN and its prokaryotic orthologs additionally have adenosine deaminase, purine nucleoside phosphorylase, and S-methyl-5'-thioadenosine phosphorylase activity, hence, combine activities of the namesake enzymes of central purine metabolism. FAMIN enables in macrophages a purine nucleotide cycle (PNC) between adenosine and inosine monophosphate and adenylosuccinate, which consumes aspartate and releases fumarate in a manner involving fatty acid oxidation and ATP-citrate lyase activity. This macrophage PNC synchronizes mitochondrial activity with glycolysis by balancing electron transfer to mitochondria, thereby supporting glycolytic activity and promoting oxidative phosphorylation and mitochondrial H+ and phosphate recycling.
Sigma-Aldrich
Palmitic acid-13C16, 99 atom % 13C, 99% (CP)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium fumarate-2,3-13C2, 99 atom % 13C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine-15N2, 98 atom % 15N
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinic acid-13C4, 99 atom % 13C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5′-Deoxyadenosine, methylthioadenosine/S-adenosylhomocysteine (MTA/SAH) nucleosidase substrate
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hypoxanthine, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
S-(5′-Adenosyl)-L-homocysteine, crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
S-(5′-Adenosyl)-L-methionine chloride dihydrochloride, ≥75%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Uridine, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5′-Deoxy-5′-(methylthio)adenosine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TEV Protease
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cholesterol Oxidase from Streptomyces sp., lyophilized powder, ≥20 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Adenosine, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Antimycin A from Streptomyces sp.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Rotenone, ≥95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2′-Deoxyadenosine monohydrate, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-(−)-Malic acid, suitable for cell culture, BioReagent, suitable for insect cell culture
SKU
Pack Size
Availability
Price
Quantity
USP
Ferulic acid, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-Glucose-13C6, ≥99 atom % 13C, ≥99% (CP)
SKU
Pack Size
Availability
Price
Quantity
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial