Skip to Content
Merck
All Photos(5)

Key Documents

D5011

Sigma-Aldrich

5′-Deoxy-5′-(methylthio)adenosine

Synonym(s):

Methylthioadenosine, MTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N5O3S
CAS Number:
Molecular Weight:
297.33
Beilstein:
42420
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMF: 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CSC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23

InChI

1S/C11H15N5O3S/c1-20-2-5-7(17)8(18)11(19-5)16-4-15-6-9(12)13-3-14-10(6)16/h3-5,7-8,11,17-18H,2H2,1H3,(H2,12,13,14)/t5-,7-,8-,11-/m1/s1

InChI key

WUUGFSXJNOTRMR-IOSLPCCCSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

5′-Deoxy-5′-(methylthio)adenosine has been used as a protein methylation inhibitor to reduce E2F transcription factor 1 (E2F1) protein abundance in hepatocellular carcinoma (HCC) cells. It has also been used to inhibit histone methylation modification and study its role in hypoxia inducible factor-1 (Hif-1) nuclear transport.

Biochem/physiol Actions

5′-Deoxy-5′-(methylthio)adenosine (Methylthioadenosine) may be used as a substrate to study the specificity and kinetics of 5′-methylthioadenosinephosphorylase (MTAP) (EC2.4.2.28), a tumor suppressor gene expressed enzyme that supports the S-adenosylmethionine (AdoMet) and methionine salvage pathways.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Histone methylation regulates Hif-1 signaling cascade in activation of hepatic stellate cells
Hong F, et al.
FEBS Open Bio, 8(3), 406-415 (2018)
SET7/9 promotes hepatocellular carcinoma progression through regulation of E2F1
Gu Y, et al.
Oncology Reports, 40(4), 1863-1874 (2018)
Jiwoung Chung et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(29), 12089-12094 (2011-07-07)
Quorum sensing (QS) controls certain behaviors of bacteria in response to population density. In gram-negative bacteria, QS is often mediated by N-acyl-L-homoserine lactones (acyl-HSLs). Because QS influences the virulence of many pathogenic bacteria, synthetic inhibitors of acyl-HSL synthases might be
Tobias J Erb et al.
Nature chemical biology, 8(11), 926-932 (2012-10-09)
Functional assignment of uncharacterized proteins is a challenge in the era of large-scale genome sequencing. Here, we combine in extracto NMR, proteomics and transcriptomics with a newly developed (knock-out) metabolomics platform to determine a potential physiological role for a ribulose-1,5-bisphosphate
Ivan Hemeon et al.
Analytical chemistry, 83(12), 4996-5004 (2011-05-07)
DNA (cytosine-5)-methyltransferases (DNMTs) catalyze the transfer of a methyl group from S-adenosyl-L-methionine (AdoMet) to the 5-position of cytosine residues and thereby silence transcription of regulated genes. DNMTs are important epigenetic targets. However, isolated DNMTs are weak catalysts and are difficult

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service