Skip to Content
Merck
All Photos(1)

Key Documents

489468

Sigma-Aldrich

Sodium fumarate-2,3-13C2

99 atom % 13C

Synonym(s):

Fumaric acid-2,3-13C2 disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
NaO2C13CH=13CHCO2Na
CAS Number:
Molecular Weight:
162.02
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

form

solid

mp

>300 °C (lit.)

mass shift

M+2

SMILES string

[Na+].[Na+].[O-]C(=O)\[13CH]=[13CH]\C([O-])=O

InChI

1S/C4H4O4.2Na/c5-3(6)1-2-4(7)8;;/h1-2H,(H,5,6)(H,7,8);;/q;2*+1/p-2/b2-1+;;/i1+1,2+1;;

InChI key

MSJMDZAOKORVFC-CRQBDLPISA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Zaeem Cader et al.
Cell, 180(2), 278-295 (2020-01-25)
Mutations in FAMIN cause arthritis and inflammatory bowel disease in early childhood, and a common genetic variant increases the risk for Crohn's disease and leprosy. We developed an unbiased liquid chromatography-mass spectrometry screen for enzymatic activity of this orphan protein.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service