Merck
  • Synthesis of phosphatidylated-monoterpene alcohols catalyzed by phospholipase D and their antiproliferative effects on human cancer cells.
Synthesis of phosphatidylated-monoterpene alcohols catalyzed by phospholipase D and their antiproliferative effects on human cancer cells.
Bioorganic & medicinal chemistry letters (2008-06-17)
Yukihiro Yamamoto, Masashi Hosokawa, Hideyuki Kurihara, Takashi Maoka, Kazuo Miyashita
ABSTRACT
In order to prepare functional phospholipids in the medical and pharmaceutical fields, perillyl alcohol, myrtenol, and nerol were transphosphatidylated via phospholipase D in an aqueous system. The yields of phosphatidyl-perillyl alcohol, -myrtenol, and -nerol were 79 mol %, 87 mol %, and 91 mol %, respectively. The synthetic phosphatidylated monoterpenes showed a markedly antiproliferative effect on human prostate PC-3 and human leukemia HL-60 cells at 100 microM, while the free monoterpene alcohols had no effect at 400 microM.
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Supelco
Geraniol, analytical standard
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Sigma-Aldrich
(S)-(−)-Perillyl alcohol, 96%
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
Sigma-Aldrich
(S)-(−)-Perillyl alcohol, ≥95%, FG
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%