Skip to Content
Merck
All Photos(1)

Documents

48798

Supelco

Geraniol

analytical standard

Synonym(s):

trans-3,7-Dimethyl-2,6-octadien-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1722456
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.31 (vs air)

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.474 (lit.)
n20/D 1.478

bp

229-230 °C (lit.)

density

0.879 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C\CC\C(C)=C\CO

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,7,11H,4,6,8H2,1-3H3/b10-7+

InChI key

GLZPCOQZEFWAFX-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: melissa mentha thymus

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pankaj K Sharma et al.
Plant science : an international journal of experimental plant biology, 203-204, 63-73 (2013-02-19)
Plants synthesize volatile alcohol esters on environmental insult or as metabolic induction during flower/fruit development. However, essential oil plants constitutively produce them as the oil constituents. Their synthesis is catalyzed by BAHD family enzymes called alcohol acyltransferases (AATs). However, no
A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S160-S170 (2008-07-22)
A toxicologic and dermatologic review of geraniol when used as a fragrance ingredient is presented.
Erich Schmidt et al.
Natural product communications, 7(8), 1095-1098 (2012-09-18)
The essential oils of four chemotypes of Thymus vulgaris L. (Lamiaceae) were analyzed for their composition and antibacterial activity to assess their different properties. GC-MS and GC-FID analyses revealed that the essentials oils can be classified into the chemotypes thymol
Naoko Sato-Masumoto et al.
Phytochemistry, 104, 12-20 (2014-05-28)
Studies on the biosynthesis of oil compounds in Perilla will help in understanding regulatory systems of secondary metabolites and in elucidating reaction mechanisms for natural product synthesis. In this study, two types of alcohol dehydrogenases, an aldo-keto reductase (AKR) and
Jesús F Arteaga et al.
Molecules (Basel, Switzerland), 17(5), 5126-5138 (2012-05-05)
Antioxidant activity of a number of small (low molecular weight) natural compounds found in spices, condiments or drugs (gallic acid, sesamol, eugenol, thymol, carvacrol, vanillin, salicylaldehyde, limonene, geraniol, 4-hexylresorcinol, etc.) has been evaluated using electrochemical and DPPH• radical scavenging measurements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service