Merck
  • Cellular apoptosis and cytotoxicity of phenolic compounds: a quantitative structure-activity relationship study.
Cellular apoptosis and cytotoxicity of phenolic compounds: a quantitative structure-activity relationship study.
Journal of medicinal chemistry (2005-11-11)
Cynthia D Selassie, Sanjay Kapur, Rajeshwar P Verma, Melissa Rosario
ABSTRACT
In this comprehensive study on the caspase-mediated apoptosis-inducing effect of 51 substituted phenols in a murine leukemia cell line (L1210), we determined the concentrations needed to induce caspase activity by 50% (I50) and utilized these data to develop the following quantitative structure-activity relationship (QSAR) model: log 1/I50 = 1.06 B5(2) + 0.33 B5(3) - 0.18pi(2,4) - 0.92. B5(3) and B5(2) represent steric terms, while pi(2,4) represents the hydrophobic character of the substituents on the ring. The strong dependence of caspase-mediated apoptosis on mostly steric parameters suggests that the process is a receptor-mediated interaction with caspases or mitochondrial proteins being the likely targets. Conversely, cytotoxicity studies of 65 electron-releasing phenols in the L1210 cell line led to the development of the following equation: log 1/ID50 = -1.39sigma+ - 0.28 B5(2,6) + 0.16 log P - 0.58I(2) - 1.04I(1) + 3.90. The low coefficient with log P may pertain to cellular transport that may be enhanced by a modest increase in overall hydrophobicity, while the presence of sigma+ is consistent with the suggestion that radical stabilization is of prime importance in the case of electron-releasing substituents. On the other hand, the QSAR for the interactions of 27 electron-attracting phenols in L1210 cells, log 1/ID50 = 0.56 log P - 0.30 B5(2) + 2.79, suggests that hydrophobicity, as represented by log P is of critical importance. Similar cytotoxicity patterns are observed in other mammalian cell lines such as HL-60, MCF-7, CCRF-CEM, and CEM/VLB. The significant differences between the cytotoxicity and apoptosis QSAR for electron-releasing phenols suggest that cytotoxicity involves minimal apoptosis in most of these substituted monophenols.
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
2-tert-Butyl-4-methylphenol, 99%
Sigma-Aldrich
4-Propylphenol, 99%
Sigma-Aldrich
3-Aminophenol, 98%
Sigma-Aldrich
2,6-Dimethoxyphenol, 99%
Sigma-Aldrich
4-Fluorophenol, 99%
Sigma-Aldrich
2-Isopropylphenol, 98%
Sigma-Aldrich
4-Ethylphenol, 99%
Sigma-Aldrich
4-tert-Butylphenol, 99%
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
2,6-Xylenol, ≥99%, FG
Sigma-Aldrich
2-Propylphenol, 98%
Sigma-Aldrich
2,4,6-Trimethylphenol, 97%
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)
Sigma-Aldrich
2-Ethoxyphenol, 98%
Sigma-Aldrich
3-Methoxyphenol, 96%
Sigma-Aldrich
4-Chlorophenol, ≥99%
Sigma-Aldrich
4-Octylphenol, 99%
Sigma-Aldrich
4-Ethylphenol, ≥98%, FG
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Supelco
o-Cresol, analytical standard
Supelco
Diethylstilbestrol, VETRANAL®, analytical standard
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
2-Isopropylphenol, ≥98%, FG
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
2-Hydroxybenzyl alcohol, 99%
Sigma-Aldrich
4-Methylcatechol, ≥95%
Sigma-Aldrich
o-Cresol, ≥99%
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
4′-Hydroxyacetophenone, 99%