Skip to Content
Merck
All Photos(1)

Documents

328456

Sigma-Aldrich

3-Methoxyphenol

96%

Synonym(s):

3-Hydroxyanisol, Resorcinol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4OH
CAS Number:
Molecular Weight:
124.14
Beilstein:
1209898
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.552 (lit.)

bp

113-115 °C/5 mmHg (lit.)

density

1.131 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(O)c1

InChI

1S/C7H8O2/c1-9-7-4-2-3-6(8)5-7/h2-5,8H,1H3

InChI key

ASHGTJPOSUFTGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methoxyphenol was used in synthesis of:
  • C(4) symmetric calix[4]resorcinarene
  • 2-nitroso-5-methoxyphenol
  • 6-methoxy-2(3H)-benzoxazolone

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alejandro Cordero Vargas et al.
Organic letters, 5(20), 3717-3719 (2003-09-26)
[reaction: see text] A short synthesis of (+/-)-10-norparvulenone and (+/-)-O-methylasparvenone was developed starting from commercially available m-methoxyphenol, hinging on a xanthate-mediated addition-cyclization sequence for the construction of the alpha-tetralone subunit.
L G Fenoll et al.
Biophysical chemistry, 84(1), 65-76 (2000-03-21)
Tyrosinase hydroxylates 3-hydroxyanisole in the 4-position. The reaction product accumulates in the reaction medium with a lag time (tau) which diminishes with increasing concentrations of enzyme and lengthens with increasing concentrations of substrate, thus fulfilling all the predictions of the
Mario C Foti et al.
The Journal of organic chemistry, 73(6), 2408-2411 (2008-02-26)
The m-methoxy group is normally electron-withdrawing (EW), sigma(m) = +0.12, sigma(m+) = +0.05. The strong EW activity of a phenoxyl radical's O* atom causes the m-methoxy group to become electron-donating (ED), sigma(m)(+) = -0.14. In valence bond terms, this can
Improved procedures for the preparation of 2-nitro-5-methoxyphenol and 6-methoxy-2 (3H)-benzoxazolone from 3-methoxyphenol.
Maleski RJ.
Synthetic Communications, 23(3), 343-348 (1993)
McIldowie et al.
Organic letters, 2(24), 3869-3871 (2000-12-02)
The Lewis acid catalyzed condensation of 3-methoxyphenol with octanal produced the C(4) symmetric calix[4]resorcinarene 2, in high yield. Of the numerous stereo- and regioisomers possible, the rccc isomer with C(4) symmetry was the only product isolated (as a racemate). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service