Skip to Content
Merck
All Photos(1)

Key Documents

36922

Supelco

o-Cresol

analytical standard

Synonym(s):

2-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
506917
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

3.72 (vs air)

vapor pressure

0.3 mmHg ( 20 °C)

autoignition temp.

1110 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

1.47 %, 148 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

191 °C (lit.)

mp

29-31 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1ccccc1O

InChI

1S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8H,1H3

InChI key

QWVGKYWNOKOFNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Cresol is a phenol derivative and an isomer of cresol. It is commercially used as a component of cleaning agents. o-Cresol is an intermediate in the production of dyes and pharmaceuticals.[1]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

o-Cresol may be used as an analytical standard for the determination of the analyte in water samples[2][3] and transformer oil[4] by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phenol in surface waters by high-performance liquid chromatography with sorption preconcentration
Basova EM, et al.
Journal of Analytical Chemistry, 57(5), 434-439 (2002)
Removal of phenol and o-cresol by adsorption onto activated carbon
Vasu AE
Journal of Chemistry, 5(2), 224-232 (2008)
Determination of phenol, m-cresol and o-cresol in transformer oil by HPLC method
Vrsaljko D, et al.
Electric Power System Research, 93, 24-31 (2012)
María Eugenia Suarez-Ojeda et al.
Chemosphere, 66(11), 2096-2105 (2006-11-11)
This study examines the feasibility of coupling a Catalytic Wet Air Oxidation (CWAO), with activated carbon (AC) as catalyst, and an aerobic biological treatment to treat a high-strength o-cresol wastewater. Two goals are pursued: (a) To determine the effect of
G Koppen et al.
Journal of applied toxicology : JAT, 27(3), 238-246 (2007-01-18)
The present paper deals with the evaluation of a battery of genotoxicity biomarkers in healthy Flemish adolescents and their relation with common pollutants occurring in their life environment. DNA damage as reflected by the comet assay appeared to be most

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service