Merck
  • mTORC1 Activation Requires DRAM-1 by Facilitating Lysosomal Amino Acid Efflux.
mTORC1 Activation Requires DRAM-1 by Facilitating Lysosomal Amino Acid Efflux.
Molecular cell (2019-09-08)
Florian Beaumatin, Jim O'Prey, Valentin J A Barthet, Barbara Zunino, Jean-Philippe Parvy, Alexis Maximilien Bachmann, Margaret O'Prey, Elżbieta Kania, Pablo Sierra Gonzalez, Robin Macintosh, Laurence Y Lao, Colin Nixon, Jonathan Lopez, Jaclyn S Long, Stephen W G Tait, Kevin M Ryan
ABSTRACT
Sensing nutrient availability is essential for appropriate cellular growth, and mTORC1 is a major regulator of this process. Mechanisms causing mTORC1 activation are, however, complex and diverse. We report here an additional important step in the activation of mTORC1, which regulates the efflux of amino acids from lysosomes into the cytoplasm. This process requires DRAM-1, which binds the membrane carrier protein SCAMP3 and the amino acid transporters SLC1A5 and LAT1, directing them to lysosomes and permitting efficient mTORC1 activation. Consequently, we show that loss of DRAM-1 also impacts pathways regulated by mTORC1, including insulin signaling, glycemic balance, and adipocyte differentiation. Interestingly, although DRAM-1 can promote autophagy, this effect on mTORC1 is autophagy independent, and autophagy only becomes important for mTORC1 activation when DRAM-1 is deleted. These findings provide important insights into mTORC1 activation and highlight the importance of DRAM-1 in growth control, metabolic homeostasis, and differentiation.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Slc1a7
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Earle′s Balanced Salts, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human GYPC
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Troglitazone, ≥98% (HPLC)
Sigma-Aldrich
L-Leucine methyl ester hydrochloride, 98%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Millipore
ProteinA-Sepharose 4, Fast Flow from Staphylococcus aureus, aqueous ethanol suspension
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Doxycycline hyclate
Supelco
Bicinchoninic Acid solution
Sigma-Aldrich
L-Glutamic acid γ-(p-nitroanilide) hydrochloride, γ-glutamyl transpeptidase substrate
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
Sigma-Aldrich
Oil Red O solution, 0.5% in isopropanol
Sigma-Aldrich
MISSION® esiRNA, targeting human SLC7A5
Sigma-Aldrich
MISSION® esiRNA, targeting human SCAMP3
Sigma-Aldrich
Copper(II) sulfate solution, 4 % (w/v) (prepared from copper (II) sulfate pentahydrate)