Skip to Content
Merck
All Photos(1)

Documents

1667600

USP

Alpha Tocopherol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-α-Tocopherol, DL-all-rac-α-Tocopherol, Vitamin E

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Beilstein:
94012
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tocopherol

manufacturer/tradename

USP

density

0.950 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3

InChI key

GVJHHUAWPYXKBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alpha Tocopherol USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Vitamin E
  • Vitamin E Capsules
  • Vitamin E Preparation
  • Oil-Soluble Vitamins Tablets
  • Tocopherols Excipient
  • Vitamin E Polyethylene Glycol Succinate
  • Oil-Soluble Vitamins Capsules
  • Annatto Seed Oil Tocotrienols

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivia I Okereke et al.
The British journal of psychiatry : the journal of mental science, 206(4), 324-331 (2015-01-13)
Homocysteine-lowering nutrients may have preventive/ameliorative roles in depression. To test whether long-term B-vitamin/folate supplementation reduces depression risk. Participants were 4331 women (mean age 63.6 years), without prior depression, from the Women's Antioxidant and Folic Acid Cardiovascular Study - a randomised
Seung-Ju Yang et al.
European journal of pharmacology, 736, 26-34 (2014-05-07)
Thiazole derivatives are attractive candidates for drug development because they can be efficiently synthesized and are active against a number of diseases and conditions, including diabetes. In our present study, we investigated the anti-inflammatory and antioxidant properties of N-adamantyl-4-methylthiazol-2-amine (KHG26693)
Hiroyuki Kitajima et al.
Acta paediatrica (Oslo, Norway : 1992), 104(2), e82-e89 (2014-11-11)
Methods to improve the mental development of extremely low birthweight (ELBW) children are currently lacking. We assessed the effects of long-term supplementation of alpha-tocopherol on the neurological development of 259 school-aged ELBW children. Extremely low birthweight participants were divided into
Sarwat Jahan et al.
Systems biology in reproductive medicine, 60(4), 199-205 (2014-04-25)
The present study was performed to determine the effects of different antioxidants on testicular histopathology and oxidative damage induced by cadmium (Cd) in rat testis and prostate. Twenty five rats were equally divided into five groups (n = 5/group). The control group
Kirsten G Coupland et al.
Movement disorders : official journal of the Movement Disorder Society, 29(13), 1606-1614 (2014-01-01)
Parkinson's disease (PD) is a neurodegenerative disorder for which environmental factors influence disease risk and may act via an epigenetic mechanism. The microtubule-associated protein tau (MAPT) is a susceptibility gene for idiopathic PD. Methylation levels were determined by pyrosequencing of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service