Skip to Content
Merck
All Photos(3)

Key Documents

T2028

Sigma-Aldrich

(+)-δ-Tocopherol

≥90%

Synonym(s):

delta Tocopherol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
EC Number:
MDL number:
UNSPSC Code:
12352212
eCl@ss:
34058016
PubChem Substance ID:
NACRES:
NA.79

biological source

plant

Assay

≥90%

form

liquid

color

light yellow to dark orange

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2cc(O)cc(C)c2O1

InChI

1S/C27H46O2/c1-20(2)10-7-11-21(3)12-8-13-22(4)14-9-16-27(6)17-15-24-19-25(28)18-23(5)26(24)29-27/h18-22,28H,7-17H2,1-6H3/t21-,22-,27-/m1/s1

InChI key

GZIFEOYASATJEH-VHFRWLAGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(+)-δ-Tocopherol has been used as a reference material for its extraction from oil using high performance liquid chromatography (HPLC). It has also been used as a standard to quantify δ-tocopherol in soybean using reverse-phase high performance liquid chromatography.

Biochem/physiol Actions

Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by homo sapiens. Various isofroms and analogues of tocopherol have opposing and differentiated regulatory activities in vivo.
Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are fat soluble anti-oxidants that protect cell membranes from oxidative damage. δ-T is methylated at the 8-position. It has an anticancer property. δ-T is involved in proinflammatory response initiated by reactive oxygen species. It lowers lipid accumulation and promotes neuronal differentiation. δ-T also has antiangiogenic and natriuretic activities.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of oil extracted from the kernel of the fruit of cumare?s palm (Astrocaryum chambira Barret)
Ramirez-Nino M, et al.
Revista Facultad Nacional De Agronomia Medellin, 71(1), 8415-8422 (2018)
Tocopherols, tocotrienols and tocomonoenols: Many similar molecules but only one vitamin E
Azzi A
Redox Biology, 101259-101259 (2019)
delta-tocopherol is more active than alpha-or gamma-tocopherol in inhibiting lung tumorigenesis in vivo
Li GX, et al.
Cancer Prevention Research (Philadelphia, Pa.), 4(3), 404-413 (2011)
Effects of infrared treatment on tocopherols, total phenolics and antioxidant activity of soybean samples
Yalcin S and Basman A
Quality Assurance and Safety of Crops & Foods, 8(2), 273-281 (2016)
Michael W Clarke et al.
The American journal of clinical nutrition, 83(1), 95-102 (2006-01-10)
Some studies have shown potential benefit of vitamin E on platelet function, but several clinical trials failed to show improved cardiovascular outcome with alpha-tocopherol supplementation. Gamma-tocopherol, a major dietary form of vitamin E, may have protective properties different from those

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service