Skip to Content
Merck
All Photos(2)

Documents

T1539

Sigma-Aldrich

(+)-α-Tocopherol

Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g

Synonym(s):

2,5,7,8-Tetramethyl-2-(4′,8′,12′-trimethyltridecyl)-6-chromanol, 5,7,8-Trimethyltocol, D-α-Tocopherol, Vitamin E

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Beilstein:
4712525
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058016
PubChem Substance ID:
NACRES:
NA.75

biological source

vegetable oil

type

Type VI

product line

BioReagent

form

liquid (≥0.88M based on potency, density and molecular wt.)

potency

≥600 mg d-α-tocopherol per g

specific activity

≥1000 IU/g

mol wt

Mw 430.71 g/mol

concentration

≥600 mg/g

technique(s)

cell culture | insect: suitable

color

yellow-orange

refractive index

n20/D 1.505 (lit.)

bp

200-220 °C/0.1 mmHg (lit.)

solubility

chloroform: soluble 0.1 mL/mL, clear, faintly yellow to yellow

density

0.95 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21-,22-,29-/m1/s1

InChI key

GVJHHUAWPYXKBD-IEOSBIPESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Use in insect cell culture applications as an antioxidant.

Other Notes

Mixed constitutional (α, β, γ, δ) isomers.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura L Baxter et al.
Nutrients, 4(6), 467-490 (2012-07-24)
Vitamin E isoforms are essential nutrients that are widely used as dietary supplements and therapeutic agents for a variety of diseases. However, their pharmacokinetic (PK) properties remain poorly characterized, and high dosage animal studies may provide further information on their
Karl Michaëlsson et al.
The American journal of clinical nutrition, 99(1), 107-114 (2013-11-15)
A reduction in the formation of free radicals and oxidative stress might reduce the rate of bone loss and muscle wasting. The objective was to determine whether α-tocopherol intake or serum concentrations are associated with fracture risk in older women
Jen-Hao Cheng et al.
European journal of medicinal chemistry, 46(4), 1222-1231 (2011-02-25)
In an effort to develop novel antioxidant as anticancer agents, a series of xanthones were prepared. In vitro screening, the synthetic xanthones revealed significant inhibitory effects on xanthine oxidase and ABTS radical-cation scavenging activity. The selective compounds 2 and 8
Ying Yang et al.
Food chemistry, 141(1), 473-481 (2013-06-19)
Vitamin E is an essential micronutrient for humans and animals due to its antioxidant and non-antioxidant biological activities. The α-tocopherol acetate form is often used in foods and other products due to its high biological activity and chemical stability. In
Y Loetscher et al.
Poultry science, 92(11), 2938-2948 (2013-10-19)
Prevention of lipid oxidation needs special attention because a high proportion of fatty acids in broiler meat are unsaturated. A feeding experiment was conducted to evaluate the antioxidant effect of dietary addition of rosemary, chokeberry pomace, rosehip, or nettle in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service