Skip to Content
Merck
All Photos(1)

Key Documents

1643419

USP

Telmisartan

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4′[(1,4′-Dimethyl-2′-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl][1,1′-biphenyl]-2-carboxylic acid, BIBR 277

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H30N4O2
CAS Number:
Molecular Weight:
514.62
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

telmisartan

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)-c4ccccc4C(O)=O)-c5nc6ccccc6n5C

InChI

1S/C33H30N4O2/c1-4-9-30-35-31-21(2)18-24(32-34-27-12-7-8-13-28(27)36(32)3)19-29(31)37(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)33(38)39/h5-8,10-19H,4,9,20H2,1-3H3,(H,38,39)

InChI key

RMMXLENWKUUMAY-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Telmisartan USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Telmisartan is a non-peptide AT1 angiotensin receptor antagonist.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mijeong Son et al.
Clinical therapeutics, 36(8), 1147-1158 (2014-07-07)
Rosuvastatin, a 3-hydroxy-3-methylglutaryl-coenzyme A (HMG-CoA) reductase inhibitor, and telmisartan, an angiotensin receptor blocker, are commonly prescribed in combination for the treatment of dyslipidemia accompanied by hypertension. However, the nature of the pharmacokinetic interaction between the 2 drugs is not clearly
Muralikrishna Gangadharan Komala et al.
PloS one, 9(11), e108994-e108994 (2014-11-05)
Sodium glucose cotransporter 2 (SGLT2) is the main luminal glucose transporter in the kidney. SGLT2 inhibition results in glycosuria and improved glycaemic control. Drugs inhibiting this transporter have recently been approved for clinical use and have been suggested to have
Habib Eslami et al.
Neuroscience letters, 558, 31-36 (2013-11-12)
Telmisartan is an angiotensin II type 1 receptor blocker and partial agonist of peroxisome proliferator-activated receptor gamma (PPAR-γ). Here, we investigated the protective capacity of telmisartan against high glucose (HG)-elicited oxidative damage in PC12 cells. The activity of lactate dehydrogenase
Pradip N Gore et al.
Endocrine, metabolic & immune disorders drug targets, 15(1), 64-70 (2014-12-03)
The present study was planned to focus on comparative effects of telmisartan vs lisinopril on blood pressure in patients of metabolic syndrome The study was carried out on 62 patients of metabolic syndrome from Dec 2010 to Oct 2012 in
Robert W Schrier et al.
The New England journal of medicine, 371(24), 2255-2266 (2014-11-18)
Hypertension is common in autosomal dominant polycystic kidney disease (ADPKD) and is associated with increased total kidney volume, activation of the renin-angiotensin-aldosterone system, and progression of kidney disease. In this double-blind, placebo-controlled trial, we randomly assigned 558 hypertensive participants with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service