Skip to Content
Merck
All Photos(1)

Key Documents

T8949

Sigma-Aldrich

Telmisartan

≥98% (HPLC), solid

Synonym(s):

4′[(1,4′-Dimethyl-2′-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl][1,1′-biphenyl]-2-carboxylic acid, BIBR 277

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H30N4O2
CAS Number:
Molecular Weight:
514.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >5 mg/mL at 60 °C
H2O: insoluble

originator

Boehringer Ingelheim

SMILES string

CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)-c4ccccc4C(O)=O)-c5nc6ccccc6n5C

InChI

1S/C33H30N4O2/c1-4-9-30-35-31-21(2)18-24(32-34-27-12-7-8-13-28(27)36(32)3)19-29(31)37(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)33(38)39/h5-8,10-19H,4,9,20H2,1-3H3,(H,38,39)

InChI key

RMMXLENWKUUMAY-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

Application

Telmisartan has been used as an AT1 receptor antagonist to study its effects on mouse models of myocardial infarction. This study reported that telmisartan inhibited the CCN1 upregulation and reduced CCN2 levels atrial cardiomyocytes. Telmisartan has also been used to evaluate its effect on the expression of CCN1 in kidney cortex of mice subjected to myocardial infarction. Furthermore, telmisartan has been used to test its efficacy against tumor growth in mouse models of colorectal cancer.

Biochem/physiol Actions

Telmisartan is a non-peptide AT1 angiotensin receptor antagonist.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Boehringer Ingelheim. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Telmisartan is soluble in DMSO at a concentration that is greater than 5 mg/ml. It is insoluble in water.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Sharpe et al.
Drugs, 61(10), 1501-1529 (2001-09-18)
Telmisartan is an angiotensin II receptor antagonist that is highly selective for type 1 angiotensin II receptors. It was significantly more effective than placebo in large (n >100), double-blind, randomised, multicentre clinical trials in patients with mild to moderate hypertension.
Tomasz Andrzej Bonda et al.
Folia histochemica et cytobiologica, 51(1), 84-91 (2013-05-22)
Chronic heart failure often leads to worsening of the renal function. Mediators of this process include inflammatory and neuroendocrine factors. CCN1 (Cyr 61), a member of growth factor-inducible immediate early genes, which modulates inflammation and fibrogenesis, is excreted with urine
Tomasz A Bonda et al.
Folia histochemica et cytobiologica, 50(1), 99-103 (2012-04-26)
Previous studies have reported the upregulation of CCN proteins early after acute heart injury. The aim of the present work was to evaluate the expression of the CCN1 and CCN2 proteins and their regulation by angiotensin II in the atrial
Marc P Maillard et al.
The Journal of pharmacology and experimental therapeutics, 302(3), 1089-1095 (2002-08-17)
In vitro studies have shown that telmisartan is an insurmountable angiotensin II subtype-1 (AT1) receptor antagonist. Herein, the molecular basis of this insurmountable antagonism has been investigated in vitro, and the effect of telmisartan has been compared in vivo with
Maurizio Cortada et al.
Audiology & neuro-otology, 25(6), 297-308 (2020-05-06)
Telmisartan is an angiotensin II receptor blocker that has pleiotropic effects and protective properties in different cell types. Moreover, telmisartan has also shown partial agonism on the peroxisome proliferator-activated receptor γ (PPAR-γ). Auditory hair cells (HCs) express PPAR-γ, and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service