Skip to Content
Merck
All Photos(1)

Key Documents

1633007

USP

Sulfanilamide Melting Point Standard

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Sulfanilamide, p-Aminobenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4SO2NH2
CAS Number:
Molecular Weight:
172.20
Beilstein:
511852
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulfanilamide

manufacturer/tradename

USP

mp

164-166 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ccc(cc1)S(N)(=O)=O

InChI

1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)

InChI key

FDDDEECHVMSUSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfanilamide Melting Point Standard also called Sulfanilamide is an antibacterial drug.

Application

Sulfanilamide Melting Point Standard USP reference standard for use in specified quality tests and assays.

It was used in a study to demonstrate photodecomposition in aqueous solution of cutaneous photosensitizing agents with the help of spin traps 5, 5-dimethyl-1-pyrroline-1-oxide.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SPECTROSCOPIC STUDIES OF CUTANEOUS PHOTOSENSITIZING AGENTS?II. SPIN TRAPPING OF PHOTOLYSIS PRODUCTS FROM SULFANILAMIDE AND 4-AMINOBENZOIC ACID USING 5, 5-DIMETHYL-1-PYRROLINE-1-OXIDE.
Chignell, Colin F., et al.
Photochemistry and Photobiology, 34.2 , 147-156 (1981)
Mohammad Islamuddin et al.
PLoS neglected tropical diseases, 9(1), e3321-e3321 (2015-01-09)
In the absence of vaccines and limitations of currently available chemotherapy, development of safe and efficacious drugs is urgently needed for visceral leishmaniasis (VL) that is fatal, if left untreated. Earlier we reported in vitro apoptotic antileishmanial activity of n-hexane
Danielle D Carrade Holt et al.
Stem cells and development, 23(11), 1258-1265 (2014-01-21)
Mesenchymal stem cells (MSCs) are used in both human clinical trials and veterinary medicine for the treatment of inflammatory and immune-mediated diseases. MSCs modulate inflammation by decreasing the cells and products of the inflammatory response. Stimulated equine MSCs from bone
Mei-Hsiu Chen et al.
Lasers in medical science, 30(1), 263-272 (2014-09-19)
For decades, low-level laser therapy (LLLT) has widespread applications in tendon-related injuries. Although the therapeutic effect of LLLT could be explained by photostimulation of target tissue and cells, how tenocytes sense photonic energy and convert them into cascades of cellular
Maria Ruweka Fernando et al.
PloS one, 9(4), e94188-e94188 (2014-04-17)
Macrophages are important innate immune cells that are associated with two distinct phenotypes: a pro-inflammatory (or classically activated) subset with prototypic macrophage functions such as inflammatory cytokine production and bactericidal activity, and an anti-inflammatory (or alternatively activated (AAM)) subset linked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service