Skip to Content
Merck
All Photos(1)

Key Documents

PHR1095

Supelco

Caffeine Melting Point Standard

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Caffeine, 1,3,7-Trimethylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
Beilstein:
17705
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1086006

API family

caffeine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

~Approximately 237 °C
234-236.5 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CN1C(=O)N(C)c2ncn(C)c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Caffeine is mainly used as a diuretic and as a stimulant to central nervous system, heart rate, and respiration.

Application

Caffeine may be used as a reference standard for the determination of the analyte in pharmaceutical formulations by high-performance liquid chromatography and spectrophotometry.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3327 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous spectrophotometric determination of paracetamol and caffeine in tablet formulation.
Vichare V, et al.
International Journal of PharmTech Research, 2(4), 2512-2516 (2010)
Development and validation of a simple RP-HPLC method for determination of caffeine in pharmaceutical dosage forms.
Chowdhury SR, et al.
Asian Journal of Pharmaceutical Analysis, 2(1), 1-4 (2012)
Markus Fehrholz et al.
American journal of physiology. Lung cellular and molecular physiology, 306(1), L101-L109 (2013-10-29)
Factors positively influencing surfactant homeostasis in general and surfactant protein B (SP-B) expression in particular are considered of clinical importance regarding an improvement of lung function in preterm infants. The objective of this study was to identify effects of physiological
France Denoeud et al.
Science (New York, N.Y.), 345(6201), 1181-1184 (2014-09-06)
Coffee is a valuable beverage crop due to its characteristic flavor, aroma, and the stimulating effects of caffeine. We generated a high-quality draft genome of the species Coffea canephora, which displays a conserved chromosomal gene order among asterid angiosperms. Although
Pierre Ayotte et al.
Environmental health perspectives, 113(10), 1318-1324 (2005-10-06)
The Lower North Shore region of the St. Lawrence River is home to a fish-eating population that displays an unusually high body burden of several organochlorines, including polychlorinated biphenyls (PCBs) and dioxin-like compounds (DLCs). We measured biomarkers indicative of liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service