Skip to Content
Merck
All Photos(1)

Key Documents

1086006

USP

Caffeine melting point standard

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Caffeine, 1,3,7-Trimethylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
Beilstein:
17705
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

caffeine

manufacturer/tradename

USP

mp

234-236.5 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1C(=O)N(C)c2ncn(C)c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Caffeine melting point standard USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

France Denoeud et al.
Science (New York, N.Y.), 345(6201), 1181-1184 (2014-09-06)
Coffee is a valuable beverage crop due to its characteristic flavor, aroma, and the stimulating effects of caffeine. We generated a high-quality draft genome of the species Coffea canephora, which displays a conserved chromosomal gene order among asterid angiosperms. Although
Brian A Mc Ardle et al.
Circulation. Cardiovascular imaging, 7(6), 930-937 (2014-09-04)
Cardiac allograft vasculopathy is a key prognostic determinant after heart transplant. Detection and risk stratification of patients with cardiac allograft vasculopathy are problematic. Positron emission tomography using rubidium-82 allows quantification of absolute myocardial blood flow and may have utility for
R Christie et al.
Drug testing and analysis, 6(7-8), 651-657 (2013-08-01)
The discrimination of a cross section of cathinone regioisomers, sold as 'legal highs', using Raman spectroscopy, is reported here. Mephedrone and flephedrone were identified in 'legal high' products sold in Irish head shops, and their 2, 3 and 4-isomers were
Tatiana Zyryanova et al.
PloS one, 9(6), e100513-e100513 (2014-06-21)
CD97 is a widely expressed adhesion class G-protein-coupled receptor (aGPCR). Here, we investigated the presence of CD97 in normal and malignant human skeletal muscle as well as the ultrastructural and functional consequences of CD97 deficiency in mice. In normal human
PharmGKB summary: very important pharmacogene information for N-acetyltransferase 2.
Ellen M McDonagh et al.
Pharmacogenetics and genomics, 24(8), 409-425 (2014-06-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service