1589029
USP
Pyroglutamic Acid
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
L-Pyroglutamic acid, (S)-(−)-2-Pyrrolidone-5-carboxylic acid, (S)-5-Oxo-2-pyrrolidinecarboxylic acid
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
pyroglutamic acid
manufacturer/tradename
USP
mp
160-163 °C (lit.)
application(s)
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
OC(=O)[C@@H]1CCC(=O)N1
InChI
1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m0/s1
InChI key
ODHCTXKNWHHXJC-VKHMYHEASA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service