Skip to Content
Merck
All Photos(2)

Key Documents

P5960

Sigma-Aldrich

L-Pyroglutamic acid

≥97% (titration), suitable for plant cell culture, BioXtra

Synonym(s):

(S)-(−)-2-Pyrrolidone-5-carboxylic acid, (S)-5-Oxo-2-pyrrolidinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
Beilstein:
82132
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Pyroglutamic acid, BioXtra

product line

BioXtra

Assay

≥97% (titration)

form

powder

technique(s)

cell culture | plant: suitable

impurities

≤0.0005% Phosphorus (P)

ign. residue

≤0.1%

color

white

mp

160-163 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

OC(=O)[C@@H]1CCC(=O)N1

InChI

1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m0/s1

InChI key

ODHCTXKNWHHXJC-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Hui Sun et al.
Chemical communications (Cambridge, England), 47(36), 10136-10138 (2011-08-11)
The chiral N-heterocyclic carbene bearing a proximal hydroxy group derived from L-pyroglutamic acid was found to be an efficient catalyst for the [3+2] annulation of enals and isatins to give the corresponding spirocyclic oxindolo-γ-butyrolactones in good yield with good diastereo-
Franz Gruber et al.
Current biology : CB, 23(6), 507-514 (2013-03-13)
Motivation controls behavior [1]. A variety of food-related behaviors undergo motivational modulation by hunger, satiety, and other states [2-4]. Here we searched for critical satiation factors modulating approach to an odor associated with sugar reward in Drosophila melanogaster. We selectively
P J Caspers et al.
The Journal of investigative dermatology, 116(3), 434-442 (2001-03-07)
Confocal Raman spectroscopy is introduced as a noninvasive in vivo optical method to measure molecular concentration profiles in the skin. It is shown how it can be applied to determine the water concentration in the stratum corneum as a function
Teruhiro Okuyama et al.
Science (New York, N.Y.), 343(6166), 91-94 (2014-01-05)
Social familiarity affects mating preference among various vertebrates. Here, we show that visual contact of a potential mating partner before mating (visual familiarization) enhances female preference for the familiarized male, but not for an unfamiliarized male, in medaka fish. Terminal-nerve
Andrea Bednářová et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 164(1), 91-100 (2012-10-23)
Glucagon is conventionally regarded as a hormone, counter regulatory in function to insulin and plays a critical anti-hypoglycemic role by maintaining glucose homeostasis in both animals and humans. Glucagon performs this function by increasing hepatic glucose output to the blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service