Skip to Content
Merck
All Photos(1)

Documents

83160

Sigma-Aldrich

L-Pyroglutamic acid

≥99.0% (T), for peptide synthesis

Synonym(s):

(S)-(−)-2-Pyrrolidone-5-carboxylic acid, (S)-5-Oxo-2-pyrrolidinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
Beilstein:
82132
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

L-Pyroglutamic acid, ≥99.0% (T)

Assay

≥99.0% (T)

form

solid

optical activity

[α]20/D −10.5±1°, c = 5% in H2O

optical purity

enantiomeric ratio: ≥99:1 (GC)

reaction suitability

reaction type: solution phase peptide synthesis

ign. residue

≤0.05%

mp

155-162 °C
160-163 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)[C@@H]1CCC(=O)N1

InChI

1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m0/s1

InChI key

ODHCTXKNWHHXJC-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Pyroglutamic acid is used in the synthesis of:
  • Nonproteinogenic amino acids such as (3S,4R)-3,4-dimethyl-L-pyroglutamic acid and (3S,4R)-3,4-dimethyl-L-glutamine.
  • Chiral N-heterocyclic carbenes (NHCs) as catalysts for the asymmetric dimerization of alkylarylketenes to give the corresponding α-quaternary β-alkylidenyl-β-lactones.

It is also used in the total synthesis of (−)-stemoamide and celogentin C.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and analysis of the sterically constrained L-glutamine analogues (3S, 4R)-3, 4-dimethyl-L-glutamine and (3S, 4R)-3, 4-dimethyl-L-pyroglutamic acid
Acevedo CM et al.
Tetrahedron, 57(30), 6353-6359 (2001)
Asymmetric Dimerization of Disubstituted Ketenes Catalyzed by N-Heterocyclic Carbenes
Lv H and Zhang et al.
advanced synthesis and catalysis, 350(17), 2715-2718 (2008)
Total Synthesis of Celogentin C by Stereoselective C-H Activation
Feng Y and Chen G
Angewandte Chemie (International ed. in English), 122(5), 970-973 (2010)
Total Syntheses of (?)-and (−)-Stemoamide
Jacobi PA and Lee K
Journal of the American Chemical Society, 122(18), 4295-4303 (2000)
Franz Gruber et al.
Current biology : CB, 23(6), 507-514 (2013-03-13)
Motivation controls behavior [1]. A variety of food-related behaviors undergo motivational modulation by hunger, satiety, and other states [2-4]. Here we searched for critical satiation factors modulating approach to an odor associated with sugar reward in Drosophila melanogaster. We selectively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service