Skip to Content
Merck

Skip To

1531007

USP

Phenylbutazone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Butyl-1,2-diphenyl-3,5-pyrazolidinedione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H20N2O2
CAS Number:
Molecular Weight:
308.37
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

phenylbutazone

manufacturer/tradename

USP

mp

106-108 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC1C(=O)N(N(C1=O)c2ccccc2)c3ccccc3

InChI

1S/C19H20N2O2/c1-2-3-14-17-18(22)20(15-10-6-4-7-11-15)21(19(17)23)16-12-8-5-9-13-16/h4-13,17H,2-3,14H2,1H3

InChI key

VYMDGNCVAMGZFE-UHFFFAOYSA-N

Gene Information

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenylbutazone USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Phenylbutazone Boluses
  • Phenylbutazone Injection
  • Phenylbutazone Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P120000080184P8386
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, TraceCERT®

grade

-

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

manufacturer/tradename

-

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

-

API family

phenylbutazone

API family

phenylbutazone

API family

-

API family

-

Gene Information

human ... PTGS1(5742), PTGS2(5743)

Gene Information

human ... PTGS1(5742), PTGS2(5743)

Gene Information

human ... PTGS1(5742), PTGS2(5743)

Gene Information

human ... CYP2C9(1559), PTGIS(5740), PTGS1(5742), PTGS2(5743)


Still not finding the right product?

Explore all of our products under Phenylbutazone


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters