Skip to Content
Merck
All Photos(2)

Key Documents

80184

Supelco

Phenylbutazone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

4-Butyl-1,2-diphenyl-3,5-pyrazolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O2
CAS Number:
Molecular Weight:
308.37
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

106-108 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCC1C(=O)N(N(C1=O)c2ccccc2)c3ccccc3

InChI

1S/C19H20N2O2/c1-2-3-14-17-18(22)20(15-10-6-4-7-11-15)21(19(17)23)16-12-8-5-9-13-16/h4-13,17H,2-3,14H2,1H3

InChI key

VYMDGNCVAMGZFE-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Lees et al.
Veterinary journal (London, England : 1997), 196(3), 294-303 (2013-06-01)
The presence of horse meat in food products destined for human consumption and labelled as beef has raised several concerns of public interest. This review deals solely with one aspect of these concerns; samples of equine tissue from horses destined
Bone marrow depression due to mianserin, phenylbutazone, oxyphenbutazone, and chloramphenicol--Part I.
S Chaplin
Adverse drug reactions and acute poisoning reviews, 5(2), 97-136 (1986-01-01)
L R Soma et al.
Journal of veterinary pharmacology and therapeutics, 35(1), 1-12 (2011-06-15)
This review presents a brief historical prospective of the genesis of regulated medication in the US racing industry of which the nonsteroidal anti-inflammatory drug (NSAID) phenylbutazone (PBZ) is the focus. It presents some historical guideposts in the development of the
Robert D Combes
Alternatives to laboratory animals : ATLA, 41(3), 235-248 (2013-08-27)
Published in silico, in vitro, in vivo laboratory animal and human data, together with information on biotransformation and data from structure-activity analyses with two decision-tree systems (ACToR and Toxtree), have been used in a weight-of-evidence (WoE) assessment to determine whether
[Multiple manifestations of intolerance in association with anti-inflammatory agents. Role of phenylbutazone].
F Tolot et al.
Semaine des hopitaux. Therapeutique, 52(3), 185-190 (1976-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service