Skip to Content
Merck
All Photos(1)

Documents

P1200000

Phenylbutazone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Butyl-1,2-diphenyl-3,5-pyrazolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O2
CAS Number:
Molecular Weight:
308.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phenylbutazone

manufacturer/tradename

EDQM

mp

106-108 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCC1C(=O)N(N(C1=O)c2ccccc2)c3ccccc3

InChI

1S/C19H20N2O2/c1-2-3-14-17-18(22)20(15-10-6-4-7-11-15)21(19(17)23)16-12-8-5-9-13-16/h4-13,17H,2-3,14H2,1H3

InChI key

VYMDGNCVAMGZFE-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenylbutazone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L R Soma et al.
Journal of veterinary pharmacology and therapeutics, 35(1), 1-12 (2011-06-15)
This review presents a brief historical prospective of the genesis of regulated medication in the US racing industry of which the nonsteroidal anti-inflammatory drug (NSAID) phenylbutazone (PBZ) is the focus. It presents some historical guideposts in the development of the
Robert D Combes
Alternatives to laboratory animals : ATLA, 41(3), 235-248 (2013-08-27)
Published in silico, in vitro, in vivo laboratory animal and human data, together with information on biotransformation and data from structure-activity analyses with two decision-tree systems (ACToR and Toxtree), have been used in a weight-of-evidence (WoE) assessment to determine whether
J Aarbakke
Clinical pharmacokinetics, 3(5), 369-380 (1978-09-01)
More than 25 years after phenylbutazone was introduced as a non-steroidal anti-inflammatory agent, basic knowledge is still accumulating on its pharmacokinetics in man. Phenylbutazone is almost completely absorbed after oral administration. A large fraction of the drug in plasma is
Bone marrow depression due to mianserin, phenylbutazone, oxyphenbutazone, and chloramphenicol--Part II.
S Chaplin
Adverse drug reactions and acute poisoning reviews, 5(3), 181-196 (1986-01-01)
Peter Lees et al.
Veterinary journal (London, England : 1997), 196(3), 294-303 (2013-06-01)
The presence of horse meat in food products destined for human consumption and labelled as beef has raised several concerns of public interest. This review deals solely with one aspect of these concerns; samples of equine tissue from horses destined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service