Skip to Content
Merck
All Photos(1)

Key Documents

1378001

USP

Medroxyprogesterone acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Medroxyprogesterone 17-acetate, 17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 6α-Methyl-17α-acetoxyprogesterone, 6α-Methyl-17α-hydroxyprogesterone acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34O4
CAS Number:
Molecular Weight:
386.52
Beilstein:
2066112
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

medroxyprogesterone

manufacturer/tradename

USP

mp

206-207 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4(OC(C)=O)C(C)=O

InChI

1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1

InChI key

PSGAAPLEWMOORI-PEINSRQWSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Medroxyprogesterone acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Medroxyprogesterone Acetate Injectable Suspension
  • Medroxyprogesterone Acetate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J Mitsushita et al.
Gynecologic oncology, 79(1), 129-132 (2000-09-28)
Successful pregnancies after conservative progestin treatment to young women with endometrial carcinoma have recently been reported. However, it is not known for certain whether the lesion is completely eradicated in such patients. We present a case of residual endometrial carcinoma
Edith R Guilbert et al.
Contraception, 79(3), 167-177 (2009-02-03)
In the fall of 2007, the controversy about the contraceptive use of depot-medroxyprogesterone acetate (DMPA) and its potential impact on skeletal health reached the media in the province of Quebec, Canada, thereby becoming a matter of concern for the lay
Carolyn Westhoff
Contraception, 68(2), 75-87 (2003-09-05)
Depot-medroxyprogesterone acetate (Depo-Provera(R)) is a highly effective, nondaily hormonal contraceptive option that has been available in the United States for a decade, and worldwide for 40 years. Benefits and risks of hormonal therapy are often under scrutiny; however, long-term clinical
Frank Z Stanczyk et al.
The Journal of steroid biochemistry and molecular biology, 142, 30-38 (2013-12-03)
Medroxyprogesterone acetate (MPA) has been in clinical use for over 30 years, and was generally considered to be safe until the results of long-term studies of postmenopausal hormone therapy (HT) using treatment with conjugated equine estrogens (CEE) combined with MPA
Andrew M Kaunitz et al.
Contraception, 77(2), 67-76 (2008-01-30)
While depot medroxyprogesterone acetate (DMPA) is a highly effective contraceptive used by millions of women, its use is associated with bone mineral density (BMD) loss, raising concerns about long-term risk of osteoporosis and/or fractures. We conducted a systematic review of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service