Skip to Content
Merck
All Photos(1)

Documents

1469005

USP

Norethindrone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

19-Norethindrone, 17α-Ethynyl-19-nortestosterone, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one, 17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one, 19-Nor-17α-ethynyl-4-androsten-17β-ol-3-one, 19-Norethisterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26O2
CAS Number:
Molecular Weight:
298.42
Beilstein:
1915671
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

norethindrone

manufacturer/tradename

USP

mp

205-206 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@]12CCC(=O)C=C1CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@@]4(O)C#C

InChI

1S/C20H26O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,12,15-18,22H,4-11H2,2H3/t15-,16+,17+,18-,19-,20-/m0/s1

InChI key

VIKNJXKGJWUCNN-XGXHKTLJSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Norethindrone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Norethindrone Tablets
  • Norethindrone Acetate Tablets
  • Norethindrone and Mestranol Tablets
  • Norethindrone and Ethinyl Estradiol Tablets
  • Fluocinolone Acetonide Cream
  • Fluoxymesterone Tablets

Biochem/physiol Actions

19-norethindrone is an oral contraceptive involved in the inhibition of cytosolic sulfotransferases (SULT).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L R Wiseman et al.
Drugs & aging, 4(3), 238-256 (1994-03-01)
The combined transdermal estradiol/norethisterone therapeutic system is designed to deliver both estradiol and norethisterone into the systemic circulation at a constant rate for up to 4 days when affixed to the skin. Transdermal administration avoids hepatic first-pass metabolism, allowing therapeutic
Rajesh Rajput et al.
Journal of obstetrics and gynaecology Canada : JOGC = Journal d'obstetrique et gynecologie du Canada : JOGC, 30(8), 680-683 (2008-09-13)
The association between the progestin-only pill used for treatment of menstrual disorders and central venous sinus thrombosis (CVST) has rarely been reported in the literature. This report describes a case of central venous sinus thrombosis following intake of norethindrone acetate
B H Draper et al.
The Cochrane database of systematic reviews, (3)(3), CD005214-CD005214 (2006-07-21)
There are two injectable progestogen-only contraceptives (IPCs) that have been available in many countries in the world since 1983. They are both still extensively used in many developing countries, forming a large proportion of the health system's expenditure on contraception.
F Z Stanczyk et al.
Contraception, 42(1), 67-96 (1990-07-01)
There is limited information on the metabolism of levonorgestrel, norethindrone and structurally related contraceptive steroids. Both levonorgestrel and norethindrone undergo extensive reduction of the alpha, beta-unsaturated ketone in ring A. Levonorgestrel also undergoes hydroxylation at carbons 2 and 16. The
H F Taitel et al.
International journal of fertility and menopausal studies, 40(4), 207-223 (1995-07-01)
Norethindrone is utilized for numerous noncontraceptive therapies, especially during the menopause. Through an analysis of the literature we have described the pharmacological profile of norethindrone and the potential therapeutic implications of low-dose therapies, with an emphasis on endometrial pathology, climacteric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service