Skip to Content
Merck

Skip To

M1629

Medroxyprogesterone 17-acetate

synthetic (organic), ≥97% (HPLC), synthetic progestogen, powder

Synonym(s):

17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 6α-Methyl-17α-acetoxyprogesterone, 6α-Methyl-17α-hydroxyprogesterone acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H34O4
CAS Number:
Molecular Weight:
386.52
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
200-757-9
MDL number:
Beilstein/REAXYS Number:
2066112
Assay:
≥97% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Medroxyprogesterone 17-acetate, ≥97% (HPLC)

biological source

synthetic (organic)

Quality Segment

sterility

non-sterile

assay

≥97% (HPLC)

form

powder

mp

206-207 °C (lit.)

solubility

chloroform: 350 mg/mL, clear, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4(OC(C)=O)C(C)=O

InChI

1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1

InChI key

PSGAAPLEWMOORI-PEINSRQWSA-N

Gene Information

Biochem/physiol Actions

Medroxyprogesterone 17-acetate (MPA) is a synthetic progestin used as a contraceptive, in hormone replacement therapy and for the treatment of endometriosis. It is a more potent progestin that the nonacetylated form.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP221PHR1589Y0001484
form

powder

form

solid

form

-

form

-

assay

≥97% (HPLC)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

-

Quality Level

300

Quality Level

-

storage temp.

room temp

storage temp.

-

storage temp.

2-30°C

storage temp.

2-8°C

solubility

chloroform: 350 mg/mL, clear, colorless

solubility

-

solubility

-

solubility

-

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-


Still not finding the right product?

Explore all of our products under Medroxyprogesterone 17-acetate


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 4 - Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
M1629-10G04061834040965
M1629-1G04061838342478

Questions

Reviews

No rating value

Active Filters