Skip to Content
Merck
All Photos(2)

Documents

59353-U

Supelco

Discovery® C8 (5 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Synonym(s):

C8 Reversed-Phase Chromatography Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L7

product line

Discovery®

feature

endcapped

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

7.5% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

15 cm × 4.6 mm

surface area

200 m2/g

surface coverage

3.4 μmol/m2

impurities

<10 ppm metals

matrix

silica gel, high purity, spherical base material
fully porous particle

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

180 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Application

Discovery® C8 HPLC column has been used in the separation of fluoroquinolone antibiotics in tablets using reversed-phase high-performance liquid chromatography (RP-HPLC).

Features and Benefits

  • Excellent reproducibility
  • Faster separation of strongly hydrophobic analytes than C18 columns
  • Stable, low-bleed LC-MS separations
  • Exceptional peak shapes for basic and acidic compounds
  • Compatible with low organic/highly aqueous mobile phases

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rahul Palchaudhuri et al.
Cell reports, 13(9), 2027-2036 (2015-12-15)
Apoptosis is generally believed to be a process that requires several hours, in contrast to non-programmed forms of cell death that can occur in minutes. Our findings challenge the time-consuming nature of apoptosis as we describe the discovery and characterization
Laurence Conraux et al.
PloS one, 8(11), e79733-e79733 (2013-11-14)
The diagnostic of Amyotrophic lateral sclerosis (ALS) remains based on clinical and neurophysiological observations. The actual delay between the onset of the symptoms and diagnosis is about 1 year, preventing early inclusion of patients into clinical trials and early care
Eridan Orlando Pereira et al.
Biotechnology for biofuels, 6, 111-111 (2013-08-07)
Microorganisms are the most proficient decomposers in nature, using secreted enzymes in the hydrolysis of lignocellulose. As such, they present the most abundant source for discovery of new enzymes. Acremonium alcalophilum is the only known cellulolytic fungus that thrives in
T Mazzei et al.
The Journal of antimicrobial chemotherapy, 31 Suppl C, 1-9 (1993-03-01)
After the discovery of erythromycin and other natural compounds, including oleandomycin, spiramycin, josamycin and midecamycin, much research has been devoted to synthesizing derivatives or analogues with improved chemical, biological and pharmacokinetic properties. These new macrolides are semisynthetic molecules that differ
Jun-Yan Zhang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 37(21), 3268-3270 (2013-02-13)
To establish a method for determing the content of two isomers containd in Garcinia hanburyi by HPLC. Chromatographic column of SunFire (Waters) C8 (2.1 mm x 150 mm, 3.5 microm) was adopted, with acetonitrile-methanol-0.3% trifluoroacetic acid (36: 37:27) as the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service