Skip to Content
Merck
All Photos(3)

Key Documents

247596

Sigma-Aldrich

Sodium formate

ACS reagent, ≥99.0%

Synonym(s):

Formic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOONa
CAS Number:
Molecular Weight:
68.01
Beilstein:
3595134
EC Number:
MDL number:
UNSPSC Code:
12352103
eCl@ss:
39021901
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99.0%
grade:
ACS reagent
form:
powder or crystals
solubility:
water: soluble(lit.)

grade

ACS reagent

Quality Level

Assay

≥99.0%

form

powder or crystals

impurities

≤0.005% insolubles

mp

259-262 °C (lit.)

solubility

water: soluble(lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.001%

cation traces

Ca: ≤0.005%
Fe: ≤5 ppm
heavy metals (as Pb): ≤5 ppm

SMILES string

[Na+].[O-]C=O

InChI

1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

HLBBKKJFGFRGMU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium formate (HCOONa) is a sodium salt of formic acid. It is a significant hydrogen source. It is used as a buffering agent to regulate the pH.

Application

Sodium formate can be used:
  • As a hydrogen source in the selective synthesis of 4,4′-diaminostilbene-2,2′-disulfonic acid (DSD) from 4,4′-dinitrostilbene-2,2′-disulfonic acid (DNS) in presence of metal oxide supported gold catalyst.
  • As a heterogeneous nucleation promoter in the synthesis of ZIF-8 (Zeolitic imidazolate framework-8) membranes on α-alumina supports.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Crystal Structure of Sodium Formate, NaHCO21.
Zachariasen WH.
Journal of the American Chemical Society, 62(5), 1011-1013 (1940)
Organic acid-dependent iron mineral formation by a newly isolated iron-reducing bacterium, Shewanella sp. HN-41.
Lee JH, et al.
Geomicrobiology Journal, 24(1), 31-41 (2007)
Formylation of Aryl Halides with Carbon Monoxide and Sodium Formate in the Presence of Palladium Catalyst.
Okano T, et al.
Bulletin of the Chemical Society of Japan, 67(8), 2329-2332 (1994)
Wen-Yih Jeng et al.
Journal of structural biology, 173(1), 46-56 (2010-08-05)
β-glucosidases (EC 3.2.1.21) cleave β-glucosidic linkages in disaccharide or glucose-substituted molecules and play important roles in fundamental biological processes. β-Glucosidases have been widely used in agricultural, biotechnological, industrial and medical applications. In this study, a high yield expression (70-250 mg/l)
Eagleson M.
Concise Encyclopedia Chemistry, 997-997 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service