Skip to Content
Merck
All Photos(2)

Documents

57914-U

Supelco

SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS)

greener alternative

for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm

Synonym(s):

DVB/CAR/PDMS SPME Fiber Assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

material

metal alloy fiber
plain gray hub

needle size

23 ga

packaging

pkg of 1 ×  ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

30 μm (CAR/PDMS layer)
50 μm (DVB layer)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

2 cm

matrix active group

DVB/CAR/PDMS coating

application(s)

food and beverages

compatibility

for analyte group flavors (volatiles and semivolatiles, C3-20) (MW 40-275)
for use with autosampler

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS) manufactured with a flexible metal alloy used in the needle, plunger, and fiber core. The new metal alloy design includes a thicker, flexible plunger that is much less likely to kink or break, and helps to reinforce the needle especially when used in an auto-sampler with a sample agitator. Since the needle is more flexible and has a thinner wall than the standard stainless steel needle, a bevel has been placed on the needle to help it pierce septa materials more easily.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application


  • Rapid headspace solid-phase microextraction-gas chromatographic-time-of-flight mass spectrometric method for qualitative profiling of ice wine volatile fraction. II: Classification of Canadian and Czech ice wines using statistical evaluation of the data: This study utilizes Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS) SPME fibers for rapid analysis of volatile compounds in ice wine, showcasing their application in qualitative profiling and classification based on geographic origin. This highlights the utility of DVB/CAR/PDMS fibers in analytical chemistry and environmental sampling, specifically in the food industry for quality control and authenticity verification (Setkova L, Risticevic S, Pawliszyn J, 2007).

Other Notes

Appearance may vary. Darker spots may be visible on the fiber but will not affect performance.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edited by Sue Ann Scheppers Wercinski
Solid Phase Microextraction: A Practical Guide, 257-257 (1999)
Pawliszyn, Janusz
Solid Phase Microextraction: Theory and Practice, 264-264 (1997)
Pawliszyn, Janusz
Applications of Solid Phase Microextraction, 674-674 (1999)
Talon, R., Montel, M.
Applied SPME, 364-371 (1999)
Pawliszyn, Janusz
Handbook of Solid Phase Microextraction, 410-410 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service