Skip to Content
Merck
All Photos(3)

Documents

57318

Supelco

SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS)

greener alternative

df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

material

fused silica fiber
plain black hub

needle size

24 ga

packaging

pkg of 3 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

75 μm (CAR/PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

CAR/PDMS coating

application(s)

food and beverages

compatibility

(For gasses and low molecular weight compounds (MW 30 - 225).)
for use with manual holder

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

For Gases and Low Molecular Weight Compounds (MW 30-225)
Solid phase microextraction (SPME) technique is generally used for GC/MS analysis. It helps in simplifying and speeding up the sample preparation. It also reduces solvent requirement during extraction.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application

SPME fiber assembly (CAR/PDMS) was used:
  • to extract volatiles from headspace vial of cheese samples.
  • to measure dimethyl sulphide concentrations of Ulva lactuca.
  • in determination of 2-Heptanone concentrations.

Footnote

This SPME fiber assembly is also available with a Nitinol (NIT) core as Cat. No. 57904-U, providing enhanced fiber reproducibility, greater fiber durability, maintained selectivity for method consistency, and core inertness to ensure analyte stability. See more at SigmaAldrich.com/NITSPME .

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Talon, R., Montel, M.
Applied SPME, 364-371 (1999)
Pawliszyn, Janusz
Solid Phase Microextraction: Theory and Practice, 264-264 (1997)
Pawliszyn, Janusz
Handbook of Solid Phase Microextraction, 410-410 (2009)
Sue Ann Scheppers Wercinski
Solid Phase Microextraction: A Practical Guide, 257-257 (1999)
Pawliszyn, Janusz
Applications of Solid Phase Microextraction, 674-674 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service