Skip to Content
Merck
All Photos(2)

Key Documents

57304

Supelco

SPME fiber assembly polyacrylate (PA)

greener alternative

df 85 μm(PA, for use with manual holder, needle size 24 ga

Synonym(s):

SPME FIBER PK/3 (MANUAL),W/85UM POLYACRYLATE COATING

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

material

fused silica fiber
plain white hub

needle size

24 ga

packaging

pack of 3

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

85 μm (PA)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PA coating

application(s)

food and beverages

compatibility

for analyte group polar semivolatiles (MW 80-300)
for use with manual holder

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Solid phase microextraction technique can be used for GC/MS analysis. It helps in simplifying and speeds up the sample preparation. It also reduces solvent requirement for the extraction. Polyacrylate is used as stationary phase in extraction of insecticides.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application

SPME fiber assembly polyacrylate (PA) was used for manual sampling, during SPME of insecticides in water samples, chlorophenols in landfill leachates, headspace immersion extraction and for extraction of free fatty acids.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R N Pereira et al.
Journal of dairy science, 91(8), 2925-2937 (2008-07-25)
The disruption of the milk fat globule membrane can lead to an excessive accumulation of free fatty acids in milk, which is frequently associated with the appearance of rancid flavors. Solid-phase microextraction and gas chromatography techniques have been shown to
D Lambropoulou et al.
Fresenius' journal of analytical chemistry, 368(6), 616-623 (2001-03-07)
Two methods for the analysis of ten organophosphorus insecticides in natural waters using solid phase extraction disks containing C18 and SDB and solid phase microextraction fibers containing polyacrylate (PA) are developed. Bromophos ethyl, bromophos methyl, dichlofenthion, ethion, fenamiphos, fenitrothion, fenthion
Eranda Wanigasekara et al.
Analytical and bioanalytical chemistry, 396(1), 511-524 (2009-11-20)
Four new ionic liquids (IL) were prepared and bonded onto 5-microm silica particles for use as adsorbent in solid-phase microextraction (SPME). Two ILs contained styrene units that allowed for polymerization and higher carbon content of the bonded silica particles. Two
Solid phase microextraction sampling and gas chromatography/mass spectrometry for field detection of the chemical warfare agent O-ethyl S-(2-diisopropylaminoethyl) methylphosphonothiolate (VX).
Hook, Gary L., et al.
Journal of Separation Science, 26, 1091-1096 (2003)
Lihua Yang et al.
Journal of chromatography. A, 1104(1-2), 23-32 (2005-12-27)
Based on solid-phase microextraction (SPME) and on-fiber silylation, a method for simultaneous determinations of exogenous endocrine disrupting chemicals (EDCs) and endogenous steroid hormones in environmental aqueous and biological samples by gas chromatography-mass spectrometry (GC-MS) was developed. The selected target compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service