55261-U
HybridSPE®-Phospholipid solid phase extraction (SPE) Cartridge
Cartridge, bed wt. 30 mg, volume 1 mL, pk of 100
Synonym(s):
HybridSPE (phospholipid and protein removal) SPE cartridge tube, 1 mL
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41115712
NACRES:
NB.21
Recommended Products
Product Name
HybridSPE®-Phospholipid, Cartridge, bed wt. 30 mg, volume 1 mL, pk of 100
material
PE frit (20 μm)
polypropylene hardware
product line
HybridSPE®
composition
bed wt., 30 mg
packaging
pk of 100
technique(s)
solid phase extraction (SPE): suitable
volume
1 mL
matrix active group
zirconia-based phase
Looking for similar products? Visit Product Comparison Guide
General description
HybridSPE-Phospholipid technology is a simple and generic sample prep platform designed for the gross level removal of endogenous protein and phospholipid interferences from biological plasma and serum prior to LC-MS or LC-MS/MS analysis. Biological plasma or serum is first subjected to protein precipitation via the addition and mixing of acidified acetonitrile. Precipitated proteins are then removed by centrifugation and the resulting supernatant is loaded on the HybridSPE-Phospholipid 96-well plate or cartridge which acts as a chemical filter that specifically targets the removal of endogenous sample phospholipids. The phospholipid retention mechanism is based on a highly selective Lewis acid-base interaction between the proprietary zirconia ions functionally bonded to the HybridSPE-Phospholipid stationary phase and the phosphate moiety consistent with all phospholipids. The resulting eluent is ready for immediate LC-MS or LC-MS-MS analysis.
The "In-well" and "In-cartridge" precipitation methods are available for the HybridSPE-Phospholipid 96-well version and HybridSPE-Phospholipid Ultra cartridge in which biological plasma/serum is first added to either the well or cartridge, followed by acidified acetonitrile (precipitation agent). After a brief mixing/vortexing step, vacuum is applied. Because the 96-well and Ultra cartridge versions contain a series of low porosity hydrophobic filters/frits, the packed-bed filter/frit assembly acts as a depth filter facilitating the concurrent removal of both phospholipids and precipitated proteins during the extraction process. Standard HybridSPE-Phospholipid cartridges require an "off-line" precipitation method.
The "In-well" and "In-cartridge" precipitation methods are available for the HybridSPE-Phospholipid 96-well version and HybridSPE-Phospholipid Ultra cartridge in which biological plasma/serum is first added to either the well or cartridge, followed by acidified acetonitrile (precipitation agent). After a brief mixing/vortexing step, vacuum is applied. Because the 96-well and Ultra cartridge versions contain a series of low porosity hydrophobic filters/frits, the packed-bed filter/frit assembly acts as a depth filter facilitating the concurrent removal of both phospholipids and precipitated proteins during the extraction process. Standard HybridSPE-Phospholipid cartridges require an "off-line" precipitation method.
Application
- Less is more: a methodological assessment of extraction techniques for per- and polyfluoroalkyl substances (PFAS) analysis in mammalian tissues.: This study assesses various extraction methods, including HybridSPE®-Phospholipid, for analyzing PFAS in tissues, highlighting efficiency and reproducibility (Mertens et al., 2023).
- Rapid analysis of 65 pharmaceuticals and 7 personal care products in plasma and whole-body tissue samples of fish using acidic extraction, zirconia-coated silica cleanup, and liquid chromatography-tandem mass spectrometry.: This paper describes a rapid analytical method incorporating HybridSPE®-Phospholipid for sample preparation, demonstrating its application in environmental toxicology (Tanoue et al., 2020).
Features and Benefits
- For use in conjunction with off-line protein precipitation.
- Merges the simplicity of protein precipitation and the selectivity of SPE via the targeted removal of phospholipids
- Reduce ion-suppression through the complete removal of phospholipids and precipitated proteins
- 2-3 step generic procedure
- Minimal to no method development
- Available in 96-well and 1 mL cartridge dimensions
Legal Information
HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany
Application
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Junji Ohnishi et al.
Human genomics, 11(1), 21-21 (2017-09-04)
Growing evidence suggests that spiritual/religious involvement may have beneficial effects on both psychological and physical functions. However, the biological basis for this relationship remains unclear. This study explored the role of spiritual/religious involvement across a wide range of biological markers
Sara Anjos et al.
Journal of proteome research, 18(8), 3174-3183 (2019-07-11)
Obesity is a public health problem and a risk factor for pathologies such type 2 diabetes mellitus, cardiovascular diseases, and nonalcoholic fatty liver disease. Given these clinical implications, there is a growing interest to understand the pathophysiological mechanism of obesity.
Takahiko Mitsui et al.
International urology and nephrology, 52(2), 233-238 (2019-10-07)
The present study was conducted to identify metabolites using a metabolomics approach and investigate the relationship between these metabolites and urgency as a major symptom of overactive bladder (OAB). In 47 male participants without any apparent neurological disease, OAB was
Seiichiro Sakao et al.
Molecular medicine reports, 23(4) (2021-01-27)
It is generally considered that there is an increase in glycolysis in the hypertrophied right ventricle (RV) during pulmonary hypertension (PH), which leads to a decrease in glucose oxidation through the tricarboxylic acid (TCA) cycle. Although recent studies have demonstrated
Marie Bagge Jensen et al.
Food chemistry, 345, 128835-128835 (2020-12-16)
Growing evidence of vitamin K's importance in human health beyond blood coagulation and bone health necessitates its further research. A method involving extraction, lipase treatment, clean-up, and detection and quantification by LC-ESI-MS/MS of phylloquinone (PK), menaquinone-4 (MK-4), menaquinone-7 (MK-7) and
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service