Skip to Content
Merck
All Photos(1)

Documents

MAK122

Sigma-Aldrich

Phospholipid Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Phospholipid Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

application(s)

cosmetics
food and beverages
pharmaceutical

detection method

colorimetric
fluorometric

shipped in

dry ice

storage temp.

−20°C

General description

Phospholipids are a class of lipids which constitute a major component of cell membranes and play important roles in signal transduction. Most phospholipids contain one diglyceride, a phosphate group, and one choline.

In this assay, phospholipids (such as lecithin, lysolecithin and sphingomyelin) are enzymatically hydrolyzed to choline which is determined using choline oxidase and a H2O2-specific dye. This results in a colorimetric (570 nm)/ fluorometric (λex = 530/λem = 585 nm) product directly proportional to the phospholipid concentration in the sample. The range of linear detection is 3 - 200 μM for colorimetric assays and 0.6 - 20 μM for fluorometric assays.

Application

Phospholipid assay kit has been used in biochemical analysis and phospholipid quantitation.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the detection of phospholipids in biological samples

Principle

In this assay, phospholipids (such as lecithin, lysolecithin and sphingomyelin) are enzymatically hydrolyzed to choline which is determined using choline oxidase and a H2O2-specific dye. This results in a colorimetric (570 nm)/ fluorometric (λex = 530/λem = 585 nm) product directly proportional to the phospholipid concentration in the sample. The range of linear detection is 3 - 200 μM for colorimetric assays and 0.6 - 20 μM for fluorometric assays.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin M Tharp et al.
American journal of physiology. Gastrointestinal and liver physiology, 310(10), G855-G864 (2016-04-02)
Gallstone disease is a widespread disorder costing billions for annual treatment in the United States. The primary mechanisms underlying gallstone formation are biliary cholesterol supersaturation and gallbladder hypomotility. The relative contribution of these two processes has been difficult to dissect
Zein Shaban Ibrahim et al.
Molecular medicine reports, 13(4), 3017-3026 (2016-03-05)
The progression of diabetic nephropathy (DN) is accelerated by smoking. The current study investigated the ability of curcumin to protect the kidneys against damage from oxidative stress induced by diabetes mellitus (DM) and nicotine (NC). A total of 24 male
Stephanie Chin et al.
Cells, 8(8) (2019-08-03)
The recent cryo-electron microscopy structures of zebrafish and the human cystic fibrosis transmembrane conductance regulator (CFTR) provided unprecedented insights into putative mechanisms underlying gating of its anion channel activity. Interestingly, despite predictions based on channel activity measurements in biological membranes
Vineela Parvathaneni et al.
Pharmaceutics, 12(3) (2020-03-04)
Non-small cell lung cancer (NSCLC) is a global disorder, treatment options for which remain limited with resistance development by cancer cells and off-target events being major roadblocks for current therapies. The discovery of new drug molecules remains time-consuming, expensive, and
Zhizhou Kuang et al.
Virulence, 11(1), 1090-1107 (2020-08-28)
The virulence behaviors of many Gram-negative bacterial pathogens are governed by quorum-sensing (QS), a hierarchical system of gene regulation that relies on population density by producing and detecting extracellular signaling molecules. Although extensively studied under in vitro conditions, adaptation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service