Skip to Content
Merck
All Photos(1)

Key Documents

500623

Supelco

ASTM® D5442 Qualitative Retention Time Mix

6.25 % (w/w) each component, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

ASTM® D5442

CofA

current certificate can be downloaded

quality

Used to establish retention time from C12 - C60

packaging

pkg of 500 mg

concentration

6.25 % (w/w) each component

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Related Categories

General description

ANALYSIS OF PETROLEUM Waxes BY GC - Qualitative and quantitative mixes of n-paraffins are used for determining column resolution, retention times, and response factors.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Docosane
    Dodecane
    Dotriacontane
    Eicosane
    Hexacosane
    Hexadecane
    Hexatriacontane
    Octacosane
    Octadecane
    Pentacontane
    Tetracontane
    Tetracosane
    Tetradecane
    Tetratetracontane
    Triacontane
    See All (15)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service