Skip to Content
Merck
All Photos(1)

Key Documents

49452-U

Supelco

C7 - C40 Saturated Alkanes Standard

certified reference material, 1000 μg/mL each component in hexane

Synonym(s):

Saturated Alkane Calibration Mix, C7 - C40

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-8°C

General description

Quantitative standard suitable for use as a retention index marker probe.

Application

Essential Oil Analysis Essential oils are complex mixtures of chemical compounds produced by living organisms and isolated from specific plant parts of known taxonomic origin. Their composition mainly includes monoterpenes, sesquiterpenes, and their oxygenated derivatives, as well as aldehydes, alcohols, and esters. In collaboration with Chromaleont and Shimadzu, our integrated solutions enhance efficiency and sustainability for the analysis of essential oils, addressing demanding workflows through innovative research, case studies, consumables, and applications.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    Decane
    Docosane
    Dodecane
    Dotriacontane
    Eicosane
    Heneicosane
    Hentriacontane
    Heptacosane
    Heptadecane
    Heptane
    Heptatriacontane
    Hexacosane
    Hexadecane
    Hexatriacontane
    Nonacosane
    Nonadecane
    Nonane
    Nonatriacontane
    Octacosane
    Octadecane
    Octane
    Octatriacontane
    Pentacosane
    Pentadecane
    Pentatriacontane
    Tetracontane
    Tetracosane
    Tetradecane
    Tetratriacontane
    Triacontane
    Tricosane
    Tridecane
    Tritriacontane
    Undecane
    See All (34)

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ling Bai et al.
Journal of separation science, 41(21), 4031-4037 (2018-08-12)
Quantification of complex carbon-containing mixtures is typically a very time-intensive task with regards to the calibration process. A gas chromatograph with a flame ionization detector yields strong responses to organic compounds and provides a wide linear range over many orders
Qingxing Shi et al.
Environmental entomology, 46(6), 1374-1380 (2017-11-04)
Volatile compounds in Tapinoma melanocephalum (Fabricius) (Hymenoptera: Formicidae, Dolichoderinae) workers were analyzed using gas chromatography-mass spectrometry and electroantennogram responses of workers to these compounds were investigated using coupled gas chromatography-electroantennographic detection. Among 18 detected compounds, six elicited electroantennogram response, including
Cintia Akemi Oi et al.
Insects, 11(5) (2020-05-02)
Sphecophaga vesparum often parasitizes nests of vespid wasps such as Vespula vulgaris and Vespula germanica. Inside the colonies, the ectoparasitic larvae feed on the immature forms of the wasps. There are two adult forms of S. vesparum. The large, winged
Phytochemical Characterization of Rhus coriaria L. Extracts by Headspace Solid-Phase Micro Extraction Gas Chromatography, Comprehensive Two-Dimensional Liquid Chromatography, and Antioxidant Activity Evaluation
Arena, K, et al.
Molecules (Basel), 27(5), 1727-1727 (2022)
Fuzhen Liu et al.
Chemosphere, 249, 126176-126176 (2020-02-23)
Sequential soil washing and electrochemical advanced oxidation processes (EAOPs) were applied for the remediation of synthetic soil contaminated with diesel. The surfactant Tween 80 was used to enhance the extraction of diesel from synthetic soil, and diesel extraction efficiency was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service