Skip to Content
Merck
All Photos(1)

Key Documents

48519

Supelco

4-Chloro-3-methylphenol

analytical standard

Synonym(s):

4-Chloro-m-cresol, PCMC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)OH
CAS Number:
Molecular Weight:
142.58
Beilstein:
1237629
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

235 °C (lit.)

mp

63-65 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Cc1cc(O)ccc1Cl

InChI

1S/C7H7ClO/c1-5-4-6(9)2-3-7(5)8/h2-4,9H,1H3

InChI key

CFKMVGJGLGKFKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-3-methylphenol is a biocide, which is colorless crystalline compound. It can be commonly used as a preservative in pharmaceutical products and as an additive to building materials and metal-working fluids.

Application

4-Chloro-3-methylphenol may be used as an analytical standard for the determination of the analyte in water and soil samples by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-phase microextraction of the antifouling Irgarol 1051 and the fungicides dichlofluanid and 4-chloro-3-methylphenol in water samples
Penalver A, et al.
Journal of Chromatography A, 839(1-2), 253-260 (1999)
Development of SPE/HPLC-DAD to Determine Residues of Selected Disinfectant Agents in Surface Water.
Baranowska I and Wojciechowska I
Polish Journal of Environmental Studies, 21(2) (2012)
A Peñalver et al.
Journal of chromatography. A, 839(1-2), 253-260 (1999-05-18)
Three pesticides usually added to paint formulations, Irgarol 1051, dichlofluanid and 4-chloro-3-methylphenol, were determined by solid-phase microextraction (SPME) with 85-micron polyacrylate fibers and gas chromatography-mass spectrometry. The parameters affecting the SPME process (the pH, the addition of salt to the
Determination of chlorophenols in soils by a method involving alkaline extraction and solid-phase preconcentration prior to high-performance liquid chromatography
Frebortova, J
Bioscience, Biotechnology, and Biochemistry, 59(10), 1930-1932 (1995)
Fraise.PA
Russell, Hugo and Ayliffe's Principles and Practice of Disinfection, Preservation and Sterilization, (5) (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service